Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21656857-21657086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD3
Entrez Gene ID
171586
Gene Name
abhydrolase domain containing 3
Gene Aliases
LABH3
UniGene
Hs.397978
Gene Chromosome Location
Chr.18: 21650897-21704805 on Build GRCh38
Transcripts
NM_001308256.1 NM_001308257.1 NM_138340.4 XM_011525847.2 XM_017025571.1 XM_017025572.1 XM_017025573.1 XM_017025574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015549 c.1036G>C p.D346H Chr.18: 21656882-21656882 on Build GRCh38
6148173 c.935T>C p.M312T Chr.18: 21656983-21656983 on Build GRCh38
6465326 c.981G>A p.P327P Chr.18: 21656937-21656937 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546290809 Chr.18: 21656955-21656955 on Build GRCh38
rs746937180 Chr.18: 21657050-21657050 on Build GRCh38
rs889984726 Chr.18: 21656989-21656989 on Build GRCh38
rs148684195 Chr.18: 21656999-21656999 on Build GRCh38
rs753077350 Chr.18: 21656939-21656939 on Build GRCh38
rs369417846 Chr.18: 21657041-21657041 on Build GRCh38
rs781362823 Chr.18: 21656976-21656976 on Build GRCh38
rs377682556 Chr.18: 21656937-21656937 on Build GRCh38
rs752095443 Chr.18: 21656960-21656960 on Build GRCh38
rs780352760 Chr.18: 21656995-21656995 on Build GRCh38
rs748020036 Chr.18: 21656997-21656997 on Build GRCh38
rs527464163 Chr.18: 21657053-21657053 on Build GRCh38
rs556581913 Chr.18: 21656910-21656910 on Build GRCh38
rs370949749 Chr.18: 21656938-21656938 on Build GRCh38
rs750420770 Chr.18: 21656978-21656978 on Build GRCh38
rs764576277 Chr.18: 21656957-21656957 on Build GRCh38
rs944215813 Chr.18: 21657032-21657033 on Build GRCh38
rs759938686 Chr.18: 21657047-21657047 on Build GRCh38
rs981836344 Chr.18: 21656943-21656943 on Build GRCh38
rs1023577853 Chr.18: 21656923-21656923 on Build GRCh38
rs756417996 Chr.18: 21656987-21656987 on Build GRCh38
rs776188758 Chr.18: 21656918-21656918 on Build GRCh38
rs752680193 Chr.18: 21657012-21657012 on Build GRCh38
rs145138728 Chr.18: 21656962-21656962 on Build GRCh38
rs376277901 Chr.18: 21657019-21657019 on Build GRCh38

View Full Product Details