Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48955407-48955674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAX
Entrez Gene ID
581
Gene Name
BCL2 associated X, apoptosis regulator
Gene Aliases
BCL2L4
UniGene
Hs.624291
Gene Chromosome Location
Chr.19: 48954825-48961798 on Build GRCh38
Transcripts
NM_001291428.1 NM_001291429.1 NM_001291430.1 NM_001291431.1 NM_004324.3 NM_138761.3 NM_138763.3 NM_138764.4 NR_027882.1 XM_017027077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210553 c.86+2T>A p.? Chr.19: 48955601-48955601 on Build GRCh38
7325069 c.86G>T p.G29V Chr.19: 48955599-48955599 on Build GRCh38
7325070 c.86G>T p.G29V Chr.19: 48955599-48955599 on Build GRCh38
5015637 c.35-2A>G p.? Chr.19: 48955546-48955546 on Build GRCh38
6210554 c.86+2T>A p.? Chr.19: 48955601-48955601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111983025 Chr.19: 48955602-48955602 on Build GRCh38
rs950323788 Chr.19: 48955580-48955580 on Build GRCh38
rs151036634 Chr.19: 48955556-48955556 on Build GRCh38
rs779255459 Chr.19: 48955646-48955646 on Build GRCh38
rs201536708 Chr.19: 48955649-48955649 on Build GRCh38
rs759391156 Chr.19: 48955429-48955429 on Build GRCh38
rs769020310 Chr.19: 48955529-48955529 on Build GRCh38
rs934186251 Chr.19: 48955628-48955628 on Build GRCh38
rs761778071 Chr.19: 48955592-48955593 on Build GRCh38
rs747305595 Chr.19: 48955528-48955528 on Build GRCh38
rs377156983 Chr.19: 48955642-48955642 on Build GRCh38
rs757323608 Chr.19: 48955507-48955507 on Build GRCh38
rs747964813 Chr.19: 48955541-48955541 on Build GRCh38
rs771938966 Chr.19: 48955579-48955580 on Build GRCh38
rs778828909 Chr.19: 48955508-48955508 on Build GRCh38
rs368357272 Chr.19: 48955500-48955500 on Build GRCh38
rs1805416 Chr.19: 48955489-48955489 on Build GRCh38
rs575244514 Chr.19: 48955526-48955526 on Build GRCh38
rs752580867 Chr.19: 48955604-48955604 on Build GRCh38
rs767362103 Chr.19: 48955512-48955512 on Build GRCh38
rs780479031 Chr.19: 48955653-48955653 on Build GRCh38
rs200950226 Chr.19: 48955650-48955650 on Build GRCh38
rs1013958516 Chr.19: 48955509-48955509 on Build GRCh38
rs1805417 Chr.19: 48955469-48955469 on Build GRCh38
rs140986746 Chr.19: 48955582-48955582 on Build GRCh38
rs377248401 Chr.19: 48955513-48955513 on Build GRCh38
rs869190728 Chr.19: 48955550-48955551 on Build GRCh38
rs776216778 Chr.19: 48955536-48955536 on Build GRCh38
rs751559000 Chr.19: 48955576-48955576 on Build GRCh38
rs767455891 Chr.19: 48955599-48955599 on Build GRCh38
rs772446633 Chr.19: 48955520-48955520 on Build GRCh38
rs374123839 Chr.19: 48955569-48955569 on Build GRCh38
rs773594332 Chr.19: 48955586-48955587 on Build GRCh38
rs113530124 Chr.19: 48955600-48955600 on Build GRCh38
rs144179827 Chr.19: 48955554-48955554 on Build GRCh38
rs142786790 Chr.19: 48955468-48955468 on Build GRCh38
rs1001083041 Chr.19: 48955480-48955480 on Build GRCh38
rs200327680 Chr.19: 48955549-48955549 on Build GRCh38
rs1001421827 Chr.19: 48955456-48955457 on Build GRCh38
rs900281740 Chr.19: 48955501-48955501 on Build GRCh38
rs754140337 Chr.19: 48955506-48955506 on Build GRCh38
rs762579313 Chr.19: 48955560-48955560 on Build GRCh38

View Full Product Details