Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101921099-101921372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
UniGene
Hs.525634
Gene Chromosome Location
Chr.12: 101877327-101923623 on Build GRCh38
Transcripts
NM_018370.2 XM_005269004.2 XM_005269005.2 XM_017019578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7120111 c.355-1G>C p.? Chr.12: 101921215-101921215 on Build GRCh38
1510897 c.704A>G p.N235S Chr.12: 101921247-101921247 on Build GRCh38
934072 c.705T>C p.N235N Chr.12: 101921248-101921248 on Build GRCh38
1677292 c.679A>C p.T227P Chr.12: 101921222-101921222 on Build GRCh38
79755 c.363C>A p.T121T Chr.12: 101921224-101921224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745800620 Chr.12: 101921197-101921197 on Build GRCh38
rs893623120 Chr.12: 101921339-101921339 on Build GRCh38
rs909162832 Chr.12: 101921327-101921327 on Build GRCh38
rs201524018 Chr.12: 101921275-101921275 on Build GRCh38
rs757029556 Chr.12: 101921182-101921182 on Build GRCh38
rs775817310 Chr.12: 101921179-101921181 on Build GRCh38
rs776905773 Chr.12: 101921211-101921211 on Build GRCh38
rs773426983 Chr.12: 101921246-101921246 on Build GRCh38
rs946601469 Chr.12: 101921345-101921345 on Build GRCh38
rs558882331 Chr.12: 101921227-101921227 on Build GRCh38
rs575251291 Chr.12: 101921257-101921257 on Build GRCh38
rs375583636 Chr.12: 101921217-101921217 on Build GRCh38
rs201266155 Chr.12: 101921265-101921265 on Build GRCh38
rs753468674 Chr.12: 101921295-101921295 on Build GRCh38
rs199920064 Chr.12: 101921268-101921268 on Build GRCh38
rs61748066 Chr.12: 101921247-101921247 on Build GRCh38
rs187106257 Chr.12: 101921169-101921169 on Build GRCh38
rs371961267 Chr.12: 101921204-101921204 on Build GRCh38
rs975835283 Chr.12: 101921202-101921202 on Build GRCh38
rs772553241 Chr.12: 101921171-101921174 on Build GRCh38
rs74465396 Chr.12: 101921163-101921163 on Build GRCh38
rs375376130 Chr.12: 101921294-101921294 on Build GRCh38
rs1033237034 Chr.12: 101921131-101921131 on Build GRCh38
rs572676043 Chr.12: 101921203-101921203 on Build GRCh38
rs757014273 Chr.12: 101921301-101921301 on Build GRCh38
rs998787846 Chr.12: 101921127-101921127 on Build GRCh38
rs201870968 Chr.12: 101921261-101921261 on Build GRCh38
rs750748130 Chr.12: 101921234-101921234 on Build GRCh38
rs367791024 Chr.12: 101921160-101921160 on Build GRCh38
rs1055089760 Chr.12: 101921318-101921318 on Build GRCh38
rs770177193 Chr.12: 101921229-101921229 on Build GRCh38
rs374955800 Chr.12: 101921164-101921164 on Build GRCh38
rs755744641 Chr.12: 101921292-101921292 on Build GRCh38
rs372311794 Chr.12: 101921248-101921248 on Build GRCh38
rs778872437 Chr.12: 101921195-101921195 on Build GRCh38
rs760093804 Chr.12: 101921249-101921249 on Build GRCh38

View Full Product Details