Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111234824-111235097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
UniGene
Hs.514840
Gene Chromosome Location
Chr.1: 111227659-111243440 on Build GRCh38
Transcripts
NM_001025197.1 NM_001025199.1 NM_004000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1732857 c.367G>T p.E123* Chr.1: 111234944-111234944 on Build GRCh38
6482634 c.346G>C p.D116H Chr.1: 111234923-111234923 on Build GRCh38
7229871 c.358T>C p.S120P Chr.1: 111234935-111234935 on Build GRCh38
2119122 c.402C>T p.N134N Chr.1: 111234979-111234979 on Build GRCh38
4831537 c.369A>T p.E123D Chr.1: 111234946-111234946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561051920 Chr.1: 111234919-111234919 on Build GRCh38
rs781703585 Chr.1: 111235015-111235015 on Build GRCh38
rs754581807 Chr.1: 111234985-111234985 on Build GRCh38
rs1012120826 Chr.1: 111235069-111235069 on Build GRCh38
rs763681184 Chr.1: 111235052-111235076 on Build GRCh38
rs749781269 Chr.1: 111235046-111235046 on Build GRCh38
rs369945883 Chr.1: 111235035-111235035 on Build GRCh38
rs956335346 Chr.1: 111234939-111234939 on Build GRCh38
rs148892404 Chr.1: 111234907-111234907 on Build GRCh38
rs760468569 Chr.1: 111234906-111234906 on Build GRCh38
rs756097721 Chr.1: 111234967-111234967 on Build GRCh38
rs896784173 Chr.1: 111235006-111235006 on Build GRCh38
rs922803383 Chr.1: 111234952-111234952 on Build GRCh38
rs909671585 Chr.1: 111234979-111234979 on Build GRCh38
rs760098034 Chr.1: 111234914-111234914 on Build GRCh38
rs199749962 Chr.1: 111235045-111235045 on Build GRCh38
rs753775528 Chr.1: 111234983-111234983 on Build GRCh38
rs1056743191 Chr.1: 111235022-111235022 on Build GRCh38
rs1036680599 Chr.1: 111234987-111234987 on Build GRCh38
rs766184399 Chr.1: 111234977-111234977 on Build GRCh38
rs375694927 Chr.1: 111234883-111234883 on Build GRCh38
rs775664454 Chr.1: 111234930-111234930 on Build GRCh38
rs762035542 Chr.1: 111235057-111235057 on Build GRCh38
rs758061039 Chr.1: 111235001-111235001 on Build GRCh38
rs978786019 Chr.1: 111234906-111234906 on Build GRCh38
rs150717614 Chr.1: 111234938-111234938 on Build GRCh38
rs1049884526 Chr.1: 111234965-111234965 on Build GRCh38
rs35041930 Chr.1: 111234953-111234953 on Build GRCh38
rs768383261 Chr.1: 111234885-111234885 on Build GRCh38
rs757229188 Chr.1: 111235068-111235068 on Build GRCh38
rs768783077 Chr.1: 111235048-111235048 on Build GRCh38
rs748866734 Chr.1: 111234878-111234878 on Build GRCh38
rs201096719 Chr.1: 111234949-111234949 on Build GRCh38
rs895060146 Chr.1: 111235029-111235029 on Build GRCh38
rs753443223 Chr.1: 111235067-111235068 on Build GRCh38
rs764130823 Chr.1: 111234944-111234944 on Build GRCh38
rs778549810 Chr.1: 111234997-111234997 on Build GRCh38
rs200158521 Chr.1: 111234895-111234895 on Build GRCh38
rs750126695 Chr.1: 111234963-111234963 on Build GRCh38
rs532267613 Chr.1: 111235019-111235019 on Build GRCh38
rs373235167 Chr.1: 111234862-111234862 on Build GRCh38
rs767304273 Chr.1: 111234960-111234960 on Build GRCh38
rs762918253 Chr.1: 111234889-111234889 on Build GRCh38
rs374410604 Chr.1: 111235060-111235060 on Build GRCh38
rs773089831 Chr.1: 111235062-111235062 on Build GRCh38
rs774293048 Chr.1: 111234947-111234947 on Build GRCh38
rs775845730 Chr.1: 111234941-111234941 on Build GRCh38
rs779628641 Chr.1: 111234859-111234859 on Build GRCh38
rs145589851 Chr.1: 111234890-111234890 on Build GRCh38
rs183515846 Chr.1: 111235055-111235055 on Build GRCh38

View Full Product Details