Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180643947-180644220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
UniGene
Hs.712820
Gene Chromosome Location
Chr.3: 180614008-180679495 on Build GRCh38
Transcripts
NM_181426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599726 c.1901C>G p.A634G Chr.3: 180644136-180644136 on Build GRCh38
6199374 c.1880C>T p.S627L Chr.3: 180644157-180644157 on Build GRCh38
5963868 c.1597A>G p.I533V Chr.3: 180644188-180644188 on Build GRCh38
5708617 c.1817_1818AC>CG p.N606T Chr.3: 180644219-180644220 on Build GRCh38
1041667 c.1828A>C p.K610Q Chr.3: 180644209-180644209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764908281 Chr.3: 180644164-180644164 on Build GRCh38
rs988032519 Chr.3: 180644065-180644065 on Build GRCh38
rs932967013 Chr.3: 180644084-180644084 on Build GRCh38
rs863224531 Chr.3: 180644141-180644141 on Build GRCh38
rs982941733 Chr.3: 180644142-180644142 on Build GRCh38
rs956631883 Chr.3: 180644061-180644061 on Build GRCh38
rs912751506 Chr.3: 180644146-180644146 on Build GRCh38
rs992265186 Chr.3: 180644033-180644033 on Build GRCh38
rs532100779 Chr.3: 180644179-180644179 on Build GRCh38
rs768877663 Chr.3: 180644046-180644046 on Build GRCh38
rs763769524 Chr.3: 180644117-180644117 on Build GRCh38
rs753580394 Chr.3: 180644119-180644119 on Build GRCh38
rs960046353 Chr.3: 180643998-180643998 on Build GRCh38
rs776608064 Chr.3: 180644165-180644165 on Build GRCh38
rs377319302 Chr.3: 180644173-180644173 on Build GRCh38
rs576304814 Chr.3: 180644154-180644154 on Build GRCh38
rs149015275 Chr.3: 180644010-180644010 on Build GRCh38
rs559628855 Chr.3: 180644153-180644153 on Build GRCh38
rs969631070 Chr.3: 180644043-180644043 on Build GRCh38
rs373541834 Chr.3: 180644167-180644167 on Build GRCh38
rs925099974 Chr.3: 180644049-180644049 on Build GRCh38
rs371433895 Chr.3: 180644083-180644083 on Build GRCh38
rs775159331 Chr.3: 180644087-180644087 on Build GRCh38
rs370618523 Chr.3: 180644174-180644174 on Build GRCh38
rs1023016750 Chr.3: 180644026-180644026 on Build GRCh38
rs761544439 Chr.3: 180644162-180644162 on Build GRCh38
rs560658609 Chr.3: 180644115-180644115 on Build GRCh38
rs759099138 Chr.3: 180643991-180643991 on Build GRCh38

View Full Product Details