Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78180594-78180829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSBG1
Entrez Gene ID
23205
Gene Name
acyl-CoA synthetase bubblegum family member 1
Gene Aliases
BG, BG1, BGM, GR-LACS, LPD
UniGene
Hs.655760
Gene Chromosome Location
Chr.15: 78167468-78234707 on Build GRCh38
Transcripts
NM_001199377.1 NM_015162.4 XM_011521390.2 XM_011521391.2 XM_017022025.1 XM_017022026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792074 c.1249G>A p.G417S Chr.15: 78180759-78180759 on Build GRCh38
5594149 c.1181G>T p.G394V Chr.15: 78180827-78180827 on Build GRCh38
3504280 c.1211C>T p.A404V Chr.15: 78180797-78180797 on Build GRCh38
5526178 c.1193G>A p.R398Q Chr.15: 78180815-78180815 on Build GRCh38
6037783 c.1190G>A p.R397Q Chr.15: 78180818-78180818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865836208 Chr.15: 78180665-78180665 on Build GRCh38
rs981984025 Chr.15: 78180688-78180688 on Build GRCh38
rs749007684 Chr.15: 78180751-78180751 on Build GRCh38
rs761668317 Chr.15: 78180709-78180709 on Build GRCh38
rs946793111 Chr.15: 78180635-78180635 on Build GRCh38
rs544973852 Chr.15: 78180749-78180749 on Build GRCh38
rs754096251 Chr.15: 78180723-78180723 on Build GRCh38
rs17479135 Chr.15: 78180676-78180676 on Build GRCh38
rs771382400 Chr.15: 78180706-78180709 on Build GRCh38
rs193007252 Chr.15: 78180671-78180671 on Build GRCh38
rs753613833 Chr.15: 78180791-78180791 on Build GRCh38
rs1019449220 Chr.15: 78180619-78180619 on Build GRCh38
rs200646960 Chr.15: 78180759-78180759 on Build GRCh38
rs34638717 Chr.15: 78180690-78180691 on Build GRCh38
rs374008679 Chr.15: 78180739-78180739 on Build GRCh38
rs202065533 Chr.15: 78180777-78180777 on Build GRCh38
rs567144170 Chr.15: 78180649-78180649 on Build GRCh38
rs371236331 Chr.15: 78180809-78180809 on Build GRCh38
rs201408176 Chr.15: 78180719-78180719 on Build GRCh38
rs752746904 Chr.15: 78180803-78180803 on Build GRCh38
rs771462805 Chr.15: 78180745-78180745 on Build GRCh38
rs759686118 Chr.15: 78180794-78180794 on Build GRCh38
rs915367641 Chr.15: 78180621-78180621 on Build GRCh38
rs961079691 Chr.15: 78180629-78180629 on Build GRCh38
rs796851895 Chr.15: 78180680-78180680 on Build GRCh38
rs373127133 Chr.15: 78180735-78180735 on Build GRCh38
rs777970413 Chr.15: 78180811-78180811 on Build GRCh38
rs958613594 Chr.15: 78180786-78180786 on Build GRCh38
rs760571690 Chr.15: 78180783-78180783 on Build GRCh38
rs200102425 Chr.15: 78180736-78180736 on Build GRCh38
rs765463008 Chr.15: 78180802-78180802 on Build GRCh38
rs1053215613 Chr.15: 78180731-78180731 on Build GRCh38
rs202151408 Chr.15: 78180740-78180740 on Build GRCh38
rs771925659 Chr.15: 78180779-78180779 on Build GRCh38
rs574146982 Chr.15: 78180675-78180675 on Build GRCh38
rs544929691 Chr.15: 78180661-78180661 on Build GRCh38
rs761346938 Chr.15: 78180769-78180769 on Build GRCh38
rs755851172 Chr.15: 78180721-78180721 on Build GRCh38
rs772265446 Chr.15: 78180790-78180790 on Build GRCh38
rs565232292 Chr.15: 78180662-78180662 on Build GRCh38
rs755221046 Chr.15: 78180725-78180725 on Build GRCh38
rs773736059 Chr.15: 78180760-78180760 on Build GRCh38

View Full Product Details