Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49660791-49661038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBPL
Entrez Gene ID
84650
Gene Name
emopamil binding protein like
Gene Aliases
EBRP
UniGene
Hs.433278
Gene Chromosome Location
Chr.13: 49660676-49691487 on Build GRCh38
Transcripts
NM_001278636.1 NM_032565.4 NR_103802.1 NR_103803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947900 c.587A>C p.Q196P Chr.13: 49661002-49661002 on Build GRCh38
3885405 c.616C>T p.Q206* Chr.13: 49660973-49660973 on Build GRCh38
469491 c.565C>G p.L189V Chr.13: 49661024-49661024 on Build GRCh38
4391448 c.589A>G p.K197E Chr.13: 49661000-49661000 on Build GRCh38
6885864 c.606G>A p.V202V Chr.13: 49660983-49660983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372752177 Chr.13: 49660987-49660987 on Build GRCh38
rs372716512 Chr.13: 49660937-49660937 on Build GRCh38
rs562582788 Chr.13: 49660828-49660828 on Build GRCh38
rs997808228 Chr.13: 49660833-49660833 on Build GRCh38
rs755820796 Chr.13: 49660951-49660951 on Build GRCh38
rs375093022 Chr.13: 49660906-49660906 on Build GRCh38
rs376251326 Chr.13: 49660999-49660999 on Build GRCh38
rs753931178 Chr.13: 49660907-49660907 on Build GRCh38
rs746289239 Chr.13: 49660962-49660962 on Build GRCh38
rs773016953 Chr.13: 49660986-49660986 on Build GRCh38
rs532877228 Chr.13: 49660977-49660979 on Build GRCh38
rs1042932074 Chr.13: 49660850-49660850 on Build GRCh38
rs752176067 Chr.13: 49660917-49660917 on Build GRCh38
rs201080175 Chr.13: 49660946-49660946 on Build GRCh38
rs867150740 Chr.13: 49660864-49660864 on Build GRCh38
rs748948687 Chr.13: 49660966-49660966 on Build GRCh38
rs763229610 Chr.13: 49661018-49661018 on Build GRCh38
rs757770579 Chr.13: 49660928-49660928 on Build GRCh38
rs772564924 Chr.13: 49661005-49661005 on Build GRCh38
rs775630117 Chr.13: 49661008-49661008 on Build GRCh38
rs544485986 Chr.13: 49660820-49660820 on Build GRCh38
rs768214631 Chr.13: 49660969-49660969 on Build GRCh38
rs865962816 Chr.13: 49661002-49661002 on Build GRCh38
rs150496327 Chr.13: 49660981-49660981 on Build GRCh38
rs376960357 Chr.13: 49660985-49660985 on Build GRCh38
rs1000612231 Chr.13: 49660880-49660880 on Build GRCh38
rs778391226 Chr.13: 49660980-49660980 on Build GRCh38
rs779614624 Chr.13: 49660956-49660956 on Build GRCh38
rs781699048 Chr.13: 49660936-49660936 on Build GRCh38
rs760145741 Chr.13: 49660915-49660915 on Build GRCh38

View Full Product Details