Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222718908-222719167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BROX
Entrez Gene ID
148362
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
C1orf58
UniGene
Hs.534965
Gene Chromosome Location
Chr.1: 222712553-222735196 on Build GRCh38
Transcripts
NM_001288579.1 NM_001288580.1 NM_001288581.1 NM_144695.3 XM_005273065.2 XM_005273069.4 XM_006711173.3 XM_011509212.2 XM_011509213.2 XM_011509214.2 XM_017000374.1 XM_017000375.1 XM_017000376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687179 c.145G>C p.D49H Chr.1: 222718968-222718968 on Build GRCh38
1959988 c.173A>G p.K58R Chr.1: 222718996-222718996 on Build GRCh38
6541803 c.180A>T p.A60A Chr.1: 222719003-222719003 on Build GRCh38
6213725 c.102-9T>G p.? Chr.1: 222718916-222718916 on Build GRCh38
4531018 c.174G>C p.K58N Chr.1: 222718997-222718997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557135868 Chr.1: 222719068-222719068 on Build GRCh38
rs779189209 Chr.1: 222719032-222719035 on Build GRCh38
rs767260851 Chr.1: 222719071-222719071 on Build GRCh38
rs375053073 Chr.1: 222718947-222718947 on Build GRCh38
rs1013486623 Chr.1: 222718984-222718984 on Build GRCh38
rs757624267 Chr.1: 222718970-222718970 on Build GRCh38
rs903013176 Chr.1: 222719134-222719134 on Build GRCh38
rs749162851 Chr.1: 222719063-222719063 on Build GRCh38
rs758989209 Chr.1: 222719004-222719004 on Build GRCh38
rs950792559 Chr.1: 222719099-222719099 on Build GRCh38
rs779742969 Chr.1: 222719001-222719001 on Build GRCh38
rs1041944261 Chr.1: 222719132-222719132 on Build GRCh38
rs772880582 Chr.1: 222719074-222719074 on Build GRCh38
rs73124895 Chr.1: 222718940-222718940 on Build GRCh38
rs747393706 Chr.1: 222719035-222719035 on Build GRCh38
rs1044575249 Chr.1: 222718997-222718997 on Build GRCh38
rs947458557 Chr.1: 222719130-222719130 on Build GRCh38
rs570798531 Chr.1: 222718965-222718965 on Build GRCh38
rs777935159 Chr.1: 222719032-222719032 on Build GRCh38
rs746115476 Chr.1: 222719054-222719054 on Build GRCh38
rs73124896 Chr.1: 222718983-222718983 on Build GRCh38
rs773069171 Chr.1: 222719135-222719135 on Build GRCh38
rs189010024 Chr.1: 222719127-222719127 on Build GRCh38
rs553424529 Chr.1: 222718989-222718989 on Build GRCh38
rs774215997 Chr.1: 222719069-222719069 on Build GRCh38
rs939085791 Chr.1: 222718948-222718948 on Build GRCh38
rs781452943 Chr.1: 222719051-222719051 on Build GRCh38
rs568430174 Chr.1: 222718993-222718993 on Build GRCh38
rs767188717 Chr.1: 222718991-222718991 on Build GRCh38
rs1056634679 Chr.1: 222718959-222718959 on Build GRCh38
rs769820232 Chr.1: 222719056-222719056 on Build GRCh38
rs138642708 Chr.1: 222719106-222719106 on Build GRCh38
rs758120296 Chr.1: 222719070-222719070 on Build GRCh38
rs762904375 Chr.1: 222718971-222718971 on Build GRCh38
rs775371560 Chr.1: 222718963-222718963 on Build GRCh38
rs149121376 Chr.1: 222719002-222719002 on Build GRCh38
rs928646385 Chr.1: 222718987-222718987 on Build GRCh38
rs760427631 Chr.1: 222719076-222719076 on Build GRCh38

View Full Product Details