Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12813285-12813510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAND2
Entrez Gene ID
23066
Gene Name
cullin associated and neddylation dissociated 2 (putative)
Gene Aliases
TIP120B, Tp120b
UniGene
Hs.343664
Gene Chromosome Location
Chr.3: 12796654-12834818 on Build GRCh38
Transcripts
NM_001162499.1 NM_012298.2 XM_011533503.2 XM_011533504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563342 c.690G>C p.E230D Chr.3: 12813351-12813351 on Build GRCh38
4583912 c.633C>T p.C211C Chr.3: 12813294-12813294 on Build GRCh38
7310929 c.727G>T p.E243* Chr.3: 12813388-12813388 on Build GRCh38
1418688 c.934C>A p.P312T Chr.3: 12813316-12813316 on Build GRCh38
1193434 c.670G>A p.D224N Chr.3: 12813331-12813331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778825402 Chr.3: 12813402-12813402 on Build GRCh38
rs1054292603 Chr.3: 12813368-12813368 on Build GRCh38
rs561681645 Chr.3: 12813331-12813331 on Build GRCh38
rs762477317 Chr.3: 12813360-12813360 on Build GRCh38
rs907135510 Chr.3: 12813417-12813417 on Build GRCh38
rs753945557 Chr.3: 12813428-12813428 on Build GRCh38
rs761315087 Chr.3: 12813349-12813349 on Build GRCh38
rs769441474 Chr.3: 12813350-12813350 on Build GRCh38
rs369959616 Chr.3: 12813466-12813466 on Build GRCh38
rs141854255 Chr.3: 12813465-12813465 on Build GRCh38
rs961812040 Chr.3: 12813479-12813479 on Build GRCh38
rs373057597 Chr.3: 12813320-12813320 on Build GRCh38
rs751494985 Chr.3: 12813373-12813373 on Build GRCh38
rs776336567 Chr.3: 12813342-12813342 on Build GRCh38
rs772668225 Chr.3: 12813474-12813474 on Build GRCh38
rs909929844 Chr.3: 12813473-12813473 on Build GRCh38
rs941432744 Chr.3: 12813482-12813482 on Build GRCh38
rs922540314 Chr.3: 12813457-12813457 on Build GRCh38
rs749406971 Chr.3: 12813325-12813325 on Build GRCh38
rs1045647490 Chr.3: 12813412-12813412 on Build GRCh38
rs548477696 Chr.3: 12813467-12813467 on Build GRCh38
rs73139814 Chr.3: 12813384-12813384 on Build GRCh38
rs779480911 Chr.3: 12813409-12813421 on Build GRCh38
rs377242398 Chr.3: 12813330-12813330 on Build GRCh38
rs370748652 Chr.3: 12813391-12813391 on Build GRCh38
rs757451394 Chr.3: 12813431-12813431 on Build GRCh38
rs138783925 Chr.3: 12813374-12813374 on Build GRCh38
rs772670150 Chr.3: 12813359-12813359 on Build GRCh38
rs779556705 Chr.3: 12813434-12813434 on Build GRCh38
rs940285548 Chr.3: 12813316-12813316 on Build GRCh38
rs746182506 Chr.3: 12813409-12813410 on Build GRCh38
rs771516008 Chr.3: 12813326-12813326 on Build GRCh38
rs984290739 Chr.3: 12813312-12813312 on Build GRCh38
rs766285827 Chr.3: 12813372-12813372 on Build GRCh38
rs796513027 Chr.3: 12813484-12813484 on Build GRCh38
rs182712663 Chr.3: 12813336-12813336 on Build GRCh38
rs778159500 Chr.3: 12813421-12813421 on Build GRCh38
rs976622739 Chr.3: 12813439-12813439 on Build GRCh38
rs756369567 Chr.3: 12813313-12813313 on Build GRCh38

View Full Product Details