Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27963070-27963335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDX2
Entrez Gene ID
1045
Gene Name
caudal type homeobox 2
Gene Aliases
CDX-3, CDX2/AS, CDX3
UniGene
Hs.127383
Gene Chromosome Location
Chr.13: 27960918-27969368 on Build GRCh38
Transcripts
NM_001265.4 XM_011534875.2 XM_011534876.2 XM_011534877.2 XM_011534878.2 XM_011534879.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625832 c.801T>C p.P267P Chr.13: 27963256-27963256 on Build GRCh38
1477174 c.780G>A p.P260P Chr.13: 27963277-27963277 on Build GRCh38
6625831 c.781C>A p.P261T Chr.13: 27963276-27963276 on Build GRCh38
5376581 c.808C>T p.P270S Chr.13: 27963249-27963249 on Build GRCh38
6625834 c.789G>A p.P263P Chr.13: 27963268-27963268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749391107 Chr.13: 27963205-27963205 on Build GRCh38
rs753318240 Chr.13: 27963109-27963109 on Build GRCh38
rs1037761396 Chr.13: 27963108-27963108 on Build GRCh38
rs745450331 Chr.13: 27963264-27963264 on Build GRCh38
rs754828776 Chr.13: 27963122-27963122 on Build GRCh38
rs575856920 Chr.13: 27963271-27963271 on Build GRCh38
rs759257805 Chr.13: 27963254-27963274 on Build GRCh38
rs1050196 Chr.13: 27963238-27963238 on Build GRCh38
rs767763015 Chr.13: 27963189-27963189 on Build GRCh38
rs77050040 Chr.13: 27963097-27963097 on Build GRCh38
rs770693358 Chr.13: 27963232-27963232 on Build GRCh38
rs746813587 Chr.13: 27963144-27963144 on Build GRCh38
rs775147672 Chr.13: 27963163-27963163 on Build GRCh38
rs376483726 Chr.13: 27963126-27963126 on Build GRCh38
rs773511514 Chr.13: 27963140-27963141 on Build GRCh38
rs756243896 Chr.13: 27963129-27963129 on Build GRCh38
rs1021116136 Chr.13: 27963257-27963277 on Build GRCh38
rs369885345 Chr.13: 27963146-27963146 on Build GRCh38
rs142861632 Chr.13: 27963141-27963141 on Build GRCh38
rs752356804 Chr.13: 27963272-27963272 on Build GRCh38
rs767312669 Chr.13: 27963288-27963290 on Build GRCh38
rs750710720 Chr.13: 27963191-27963191 on Build GRCh38
rs544841954 Chr.13: 27963278-27963278 on Build GRCh38
rs375656067 Chr.13: 27963206-27963206 on Build GRCh38
rs1018000717 Chr.13: 27963145-27963145 on Build GRCh38
rs764455094 Chr.13: 27963113-27963113 on Build GRCh38
rs570781335 Chr.13: 27963111-27963111 on Build GRCh38
rs769586482 Chr.13: 27963268-27963268 on Build GRCh38
rs139907356 Chr.13: 27963208-27963208 on Build GRCh38
rs763499719 Chr.13: 27963164-27963164 on Build GRCh38
rs775098429 Chr.13: 27963269-27963269 on Build GRCh38
rs1805107 Chr.13: 27963180-27963180 on Build GRCh38
rs954978040 Chr.13: 27963142-27963142 on Build GRCh38
rs1805108 Chr.13: 27963105-27963105 on Build GRCh38
rs776422936 Chr.13: 27963233-27963233 on Build GRCh38
rs201546906 Chr.13: 27963143-27963143 on Build GRCh38
rs371871813 Chr.13: 27963099-27963099 on Build GRCh38
rs754918656 Chr.13: 27963207-27963207 on Build GRCh38
rs993458674 Chr.13: 27963132-27963132 on Build GRCh38
rs756333562 Chr.13: 27963192-27963192 on Build GRCh38
rs199725007 Chr.13: 27963274-27963274 on Build GRCh38
rs762233549 Chr.13: 27963290-27963290 on Build GRCh38
rs200040851 Chr.13: 27963256-27963256 on Build GRCh38
rs143217655 Chr.13: 27963280-27963281 on Build GRCh38
rs751918148 Chr.13: 27963179-27963179 on Build GRCh38
rs868051017 Chr.13: 27963155-27963155 on Build GRCh38
rs140620214 Chr.13: 27963127-27963127 on Build GRCh38
rs373555157 Chr.13: 27963176-27963176 on Build GRCh38
rs186344616 Chr.13: 27963147-27963147 on Build GRCh38
rs778816395 Chr.13: 27963133-27963133 on Build GRCh38
rs751890323 Chr.13: 27963121-27963121 on Build GRCh38
rs976477921 Chr.13: 27963168-27963168 on Build GRCh38
rs775392059 Chr.13: 27963290-27963301 on Build GRCh38
rs780049295 Chr.13: 27963196-27963196 on Build GRCh38

View Full Product Details