Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33258859-33259084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
UniGene
Hs.377484
Gene Chromosome Location
Chr.9: 33252471-33264761 on Build GRCh38
Transcripts
NM_001172415.1 NM_004323.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150313 c.745G>C p.E249Q Chr.9: 33258952-33258952 on Build GRCh38
4152249 c.696G>T p.K232N Chr.9: 33259001-33259001 on Build GRCh38
5359391 c.429G>A p.Q143Q Chr.9: 33258923-33258923 on Build GRCh38
2150333 c.675G>T p.Q225H Chr.9: 33259022-33259022 on Build GRCh38
4152250 c.351G>T p.K117N Chr.9: 33259001-33259001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746400542 Chr.9: 33258945-33258945 on Build GRCh38
rs776534914 Chr.9: 33259011-33259011 on Build GRCh38
rs773234240 Chr.9: 33258984-33258984 on Build GRCh38
rs753388761 Chr.9: 33258908-33258908 on Build GRCh38
rs140625706 Chr.9: 33259032-33259032 on Build GRCh38
rs749449480 Chr.9: 33258959-33258959 on Build GRCh38
rs945754882 Chr.9: 33258995-33258995 on Build GRCh38
rs571192965 Chr.9: 33258931-33258931 on Build GRCh38
rs780509025 Chr.9: 33258955-33258955 on Build GRCh38
rs754695010 Chr.9: 33258915-33258915 on Build GRCh38
rs748171397 Chr.9: 33258981-33258981 on Build GRCh38
rs768982200 Chr.9: 33258973-33258973 on Build GRCh38
rs568561495 Chr.9: 33259010-33259010 on Build GRCh38
rs931840979 Chr.9: 33258947-33258947 on Build GRCh38
rs377685649 Chr.9: 33258895-33258895 on Build GRCh38
rs199641418 Chr.9: 33258932-33258932 on Build GRCh38
rs987286463 Chr.9: 33258970-33258970 on Build GRCh38
rs763629554 Chr.9: 33259060-33259060 on Build GRCh38
rs775723435 Chr.9: 33259045-33259047 on Build GRCh38
rs370676610 Chr.9: 33258949-33258949 on Build GRCh38
rs760724367 Chr.9: 33258993-33258993 on Build GRCh38
rs774569452 Chr.9: 33258974-33258974 on Build GRCh38
rs547842355 Chr.9: 33259044-33259044 on Build GRCh38
rs765003937 Chr.9: 33259026-33259026 on Build GRCh38
rs983062292 Chr.9: 33259048-33259048 on Build GRCh38
rs781623410 Chr.9: 33258943-33258943 on Build GRCh38
rs766218024 Chr.9: 33258907-33258907 on Build GRCh38
rs538217810 Chr.9: 33259015-33259015 on Build GRCh38
rs772290563 Chr.9: 33258983-33258983 on Build GRCh38
rs760488919 Chr.9: 33258901-33258901 on Build GRCh38
rs374090558 Chr.9: 33259024-33259024 on Build GRCh38
rs147210903 Chr.9: 33258933-33258933 on Build GRCh38
rs752363640 Chr.9: 33259031-33259031 on Build GRCh38
rs181475436 Chr.9: 33258926-33258926 on Build GRCh38

View Full Product Details