Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71029225-71029450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCK
Entrez Gene ID
1633
Gene Name
deoxycytidine kinase
Gene Aliases
-
UniGene
Hs.709
Gene Chromosome Location
Chr.4: 70993548-71030914 on Build GRCh38
Transcripts
NM_000788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726939 c.781T>C p.*261R Chr.4: 71029376-71029376 on Build GRCh38
734799 c.779T>G p.L260W Chr.4: 71029374-71029374 on Build GRCh38
5512028 c.762A>G p.K254K Chr.4: 71029357-71029357 on Build GRCh38
7166495 c.771G>C p.L257F Chr.4: 71029366-71029366 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032248584 Chr.4: 71029358-71029358 on Build GRCh38
rs775098372 Chr.4: 71029418-71029418 on Build GRCh38
rs746729229 Chr.4: 71029376-71029376 on Build GRCh38
rs771563914 Chr.4: 71029357-71029357 on Build GRCh38
rs772852515 Chr.4: 71029368-71029368 on Build GRCh38
rs765251147 Chr.4: 71029404-71029404 on Build GRCh38
rs557664606 Chr.4: 71029306-71029306 on Build GRCh38
rs776166518 Chr.4: 71029385-71029385 on Build GRCh38
rs115663298 Chr.4: 71029286-71029286 on Build GRCh38
rs770412767 Chr.4: 71029381-71029381 on Build GRCh38
rs1002144273 Chr.4: 71029348-71029348 on Build GRCh38
rs765616732 Chr.4: 71029329-71029329 on Build GRCh38
rs748564369 Chr.4: 71029347-71029347 on Build GRCh38
rs372267150 Chr.4: 71029323-71029323 on Build GRCh38
rs749748134 Chr.4: 71029332-71029332 on Build GRCh38
rs976216211 Chr.4: 71029267-71029267 on Build GRCh38
rs1020706504 Chr.4: 71029414-71029414 on Build GRCh38
rs759359487 Chr.4: 71029393-71029393 on Build GRCh38
rs760267267 Chr.4: 71029328-71029329 on Build GRCh38
rs769259712 Chr.4: 71029341-71029341 on Build GRCh38
rs779121057 Chr.4: 71029346-71029346 on Build GRCh38
rs988190607 Chr.4: 71029410-71029410 on Build GRCh38
rs371656179 Chr.4: 71029302-71029302 on Build GRCh38
rs199819411 Chr.4: 71029322-71029322 on Build GRCh38
rs759756789 Chr.4: 71029263-71029263 on Build GRCh38
rs897863922 Chr.4: 71029265-71029265 on Build GRCh38

View Full Product Details