Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59671102-59671373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP2
Entrez Gene ID
663
Gene Name
BCL2 interacting protein 2
Gene Aliases
BNIP-2, NIP2
UniGene
Hs.592515
Gene Chromosome Location
Chr.15: 59659146-59689534 on Build GRCh38
Transcripts
NM_001320674.1 NM_001320675.1 NM_004330.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3956790 c.703A>T p.R235* Chr.15: 59671187-59671187 on Build GRCh38
3956791 c.1005A>T p.A335A Chr.15: 59671248-59671248 on Build GRCh38
3816467 c.700G>A p.D234N Chr.15: 59671190-59671190 on Build GRCh38
7251220 c.601T>C p.L201L Chr.15: 59671289-59671289 on Build GRCh38
5849467 c.702T>G p.D234E Chr.15: 59671188-59671188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759054124 Chr.15: 59671291-59671291 on Build GRCh38
rs375203496 Chr.15: 59671179-59671179 on Build GRCh38
rs773267301 Chr.15: 59671280-59671280 on Build GRCh38
rs755183480 Chr.15: 59671241-59671241 on Build GRCh38
rs371022671 Chr.15: 59671139-59671139 on Build GRCh38
rs750990056 Chr.15: 59671214-59671214 on Build GRCh38
rs146161822 Chr.15: 59671292-59671292 on Build GRCh38
rs540256199 Chr.15: 59671138-59671138 on Build GRCh38
rs780055483 Chr.15: 59671317-59671317 on Build GRCh38
rs765642936 Chr.15: 59671311-59671311 on Build GRCh38
rs766042901 Chr.15: 59671175-59671175 on Build GRCh38
rs377764966 Chr.15: 59671290-59671290 on Build GRCh38
rs757640199 Chr.15: 59671204-59671204 on Build GRCh38
rs769780337 Chr.15: 59671142-59671142 on Build GRCh38
rs993430273 Chr.15: 59671240-59671240 on Build GRCh38
rs139533727 Chr.15: 59671278-59671278 on Build GRCh38
rs770533061 Chr.15: 59671287-59671287 on Build GRCh38
rs779473072 Chr.15: 59671212-59671212 on Build GRCh38
rs930690358 Chr.15: 59671307-59671307 on Build GRCh38
rs780788843 Chr.15: 59671270-59671270 on Build GRCh38
rs769542414 Chr.15: 59671279-59671279 on Build GRCh38
rs772779513 Chr.15: 59671143-59671143 on Build GRCh38
rs1135148 Chr.15: 59671289-59671289 on Build GRCh38
rs755085567 Chr.15: 59671343-59671343 on Build GRCh38
rs751648014 Chr.15: 59671330-59671330 on Build GRCh38
rs763032040 Chr.15: 59671286-59671286 on Build GRCh38
rs768941179 Chr.15: 59671261-59671261 on Build GRCh38
rs917506742 Chr.15: 59671254-59671254 on Build GRCh38
rs558766991 Chr.15: 59671153-59671153 on Build GRCh38
rs368083466 Chr.15: 59671315-59671315 on Build GRCh38
rs543285464 Chr.15: 59671227-59671227 on Build GRCh38
rs780152765 Chr.15: 59671229-59671229 on Build GRCh38
rs149595116 Chr.15: 59671297-59671297 on Build GRCh38
rs762545974 Chr.15: 59671160-59671160 on Build GRCh38
rs762227404 Chr.15: 59671310-59671310 on Build GRCh38
rs758939010 Chr.15: 59671316-59671316 on Build GRCh38
rs754243777 Chr.15: 59671189-59671189 on Build GRCh38
rs764316738 Chr.15: 59671182-59671182 on Build GRCh38
rs758455120 Chr.15: 59671221-59671221 on Build GRCh38

View Full Product Details