Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63276441-63276686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGAP1
Entrez Gene ID
55738
Gene Name
ADP ribosylation factor GTPase activating protein 1
Gene Aliases
ARF1GAP, HRIHFB2281
UniGene
Hs.25584
Gene Chromosome Location
Chr.20: 63272785-63289793 on Build GRCh38
Transcripts
NM_001281482.1 NM_001281483.1 NM_001281484.1 NM_018209.3 NM_175609.2 XM_006723823.2 XM_011528901.1 XM_017027950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841496 c.337G>T p.D113Y Chr.20: 63276646-63276646 on Build GRCh38
7300586 c.342G>A p.K114K Chr.20: 63276651-63276651 on Build GRCh38
5041715 c.203A>T p.K68M Chr.20: 63276512-63276512 on Build GRCh38
3423810 c.206A>C p.D69A Chr.20: 63276515-63276515 on Build GRCh38
7300585 c.342G>A p.K114K Chr.20: 63276651-63276651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576715591 Chr.20: 63276573-63276573 on Build GRCh38
rs764168997 Chr.20: 63276499-63276499 on Build GRCh38
rs781685909 Chr.20: 63276477-63276478 on Build GRCh38
rs370137555 Chr.20: 63276491-63276491 on Build GRCh38
rs758957163 Chr.20: 63276584-63276584 on Build GRCh38
rs763050252 Chr.20: 63276480-63276480 on Build GRCh38
rs11542296 Chr.20: 63276630-63276630 on Build GRCh38
rs200629964 Chr.20: 63276588-63276588 on Build GRCh38
rs757459121 Chr.20: 63276559-63276559 on Build GRCh38
rs151307924 Chr.20: 63276507-63276507 on Build GRCh38
rs912197262 Chr.20: 63276613-63276613 on Build GRCh38
rs200147733 Chr.20: 63276666-63276666 on Build GRCh38
rs535336047 Chr.20: 63276469-63276469 on Build GRCh38
rs373697939 Chr.20: 63276633-63276633 on Build GRCh38
rs764140796 Chr.20: 63276658-63276658 on Build GRCh38
rs776175896 Chr.20: 63276467-63276467 on Build GRCh38
rs557202365 Chr.20: 63276589-63276589 on Build GRCh38
rs986496177 Chr.20: 63276575-63276575 on Build GRCh38
rs766387274 Chr.20: 63276487-63276487 on Build GRCh38
rs749359612 Chr.20: 63276522-63276522 on Build GRCh38
rs140491832 Chr.20: 63276590-63276590 on Build GRCh38
rs916046637 Chr.20: 63276532-63276532 on Build GRCh38
rs1024923756 Chr.20: 63276459-63276459 on Build GRCh38
rs751465768 Chr.20: 63276488-63276488 on Build GRCh38
rs116204160 Chr.20: 63276473-63276473 on Build GRCh38
rs753951040 Chr.20: 63276665-63276665 on Build GRCh38
rs768160188 Chr.20: 63276463-63276463 on Build GRCh38
rs973031358 Chr.20: 63276662-63276662 on Build GRCh38
rs776146864 Chr.20: 63276610-63276610 on Build GRCh38
rs374541250 Chr.20: 63276632-63276632 on Build GRCh38
rs774162760 Chr.20: 63276636-63276636 on Build GRCh38
rs747812610 Chr.20: 63276615-63276615 on Build GRCh38
rs769565819 Chr.20: 63276620-63276620 on Build GRCh38
rs775025548 Chr.20: 63276461-63276461 on Build GRCh38
rs759462873 Chr.20: 63276638-63276638 on Build GRCh38
rs183397013 Chr.20: 63276629-63276629 on Build GRCh38
rs190396188 Chr.20: 63276462-63276462 on Build GRCh38
rs568574915 Chr.20: 63276560-63276560 on Build GRCh38
rs765135273 Chr.20: 63276476-63276476 on Build GRCh38

View Full Product Details