Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100048729-100048968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN1
Entrez Gene ID
1369
Gene Name
carboxypeptidase N subunit 1
Gene Aliases
CPN, SCPN
UniGene
Hs.2246
Gene Chromosome Location
Chr.10: 100042308-100081885 on Build GRCh38
Transcripts
NM_001308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362480 c.1226C>T p.T409M Chr.10: 100048762-100048762 on Build GRCh38
5479489 c.1227G>A p.T409T Chr.10: 100048761-100048761 on Build GRCh38
310303 c.1230+1G>T p.? Chr.10: 100048757-100048757 on Build GRCh38
221763 c.1226C>G p.T409R Chr.10: 100048762-100048762 on Build GRCh38
7200188 c.1230+1G>A p.? Chr.10: 100048757-100048757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201305110 Chr.10: 100048810-100048810 on Build GRCh38
rs749592262 Chr.10: 100048794-100048794 on Build GRCh38
rs766856472 Chr.10: 100048871-100048871 on Build GRCh38
rs755180548 Chr.10: 100048921-100048921 on Build GRCh38
rs74152939 Chr.10: 100048937-100048937 on Build GRCh38
rs750419214 Chr.10: 100048887-100048887 on Build GRCh38
rs767386061 Chr.10: 100048875-100048875 on Build GRCh38
rs553171656 Chr.10: 100048771-100048771 on Build GRCh38
rs386747005 Chr.10: 100048937-100048938 on Build GRCh38
rs376306168 Chr.10: 100048901-100048901 on Build GRCh38
rs764912390 Chr.10: 100048756-100048756 on Build GRCh38
rs761091093 Chr.10: 100048851-100048851 on Build GRCh38
rs753988278 Chr.10: 100048906-100048906 on Build GRCh38
rs558141823 Chr.10: 100048846-100048846 on Build GRCh38
rs1046976621 Chr.10: 100048765-100048765 on Build GRCh38
rs140999407 Chr.10: 100048770-100048770 on Build GRCh38
rs745316196 Chr.10: 100048833-100048833 on Build GRCh38
rs769483937 Chr.10: 100048835-100048835 on Build GRCh38
rs878958578 Chr.10: 100048760-100048760 on Build GRCh38
rs754234111 Chr.10: 100048892-100048892 on Build GRCh38
rs545323522 Chr.10: 100048806-100048806 on Build GRCh38
rs151188411 Chr.10: 100048767-100048767 on Build GRCh38
rs779284684 Chr.10: 100048902-100048902 on Build GRCh38
rs143121841 Chr.10: 100048762-100048762 on Build GRCh38
rs1029723666 Chr.10: 100048809-100048809 on Build GRCh38
rs780243866 Chr.10: 100048789-100048789 on Build GRCh38
rs369526580 Chr.10: 100048778-100048778 on Build GRCh38
rs761626715 Chr.10: 100048860-100048860 on Build GRCh38
rs767463959 Chr.10: 100048781-100048781 on Build GRCh38
rs755269485 Chr.10: 100048801-100048801 on Build GRCh38
rs750462179 Chr.10: 100048782-100048782 on Build GRCh38
rs1031961718 Chr.10: 100048932-100048932 on Build GRCh38
rs763773688 Chr.10: 100048856-100048856 on Build GRCh38
rs115096736 Chr.10: 100048785-100048785 on Build GRCh38
rs759152678 Chr.10: 100048753-100048753 on Build GRCh38
rs372376426 Chr.10: 100048855-100048855 on Build GRCh38
rs955360034 Chr.10: 100048847-100048847 on Build GRCh38
rs1046566454 Chr.10: 100048766-100048767 on Build GRCh38
rs78940928 Chr.10: 100048938-100048938 on Build GRCh38
rs1008651337 Chr.10: 100048912-100048912 on Build GRCh38
rs913024568 Chr.10: 100048757-100048757 on Build GRCh38
rs772671251 Chr.10: 100048832-100048832 on Build GRCh38
rs138814145 Chr.10: 100048853-100048853 on Build GRCh38

View Full Product Details