Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130595408-130595671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUBP3
Entrez Gene ID
8939
Gene Name
far upstream element binding protein 3
Gene Aliases
FBP3
UniGene
Hs.673029
Gene Chromosome Location
Chr.9: 130579567-130638352 on Build GRCh38
Transcripts
NM_003934.1 XM_005272232.1 XM_006717313.2 XM_011519172.2 XM_011519174.2 XM_017015269.1 XM_017015270.1 XM_017015271.1 XM_017015272.1 XM_017015273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667730 c.164T>C p.V55A Chr.9: 130595562-130595562 on Build GRCh38
3929852 c.109C>A p.P37T Chr.9: 130595507-130595507 on Build GRCh38
4835342 c.146C>T p.S49L Chr.9: 130595544-130595544 on Build GRCh38
4391574 c.92C>G p.A31G Chr.9: 130595490-130595490 on Build GRCh38
3904784 c.131C>A p.P44H Chr.9: 130595529-130595529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018538736 Chr.9: 130595578-130595578 on Build GRCh38
rs754992820 Chr.9: 130595520-130595520 on Build GRCh38
rs759678826 Chr.9: 130595607-130595607 on Build GRCh38
rs763939144 Chr.9: 130595612-130595612 on Build GRCh38
rs774377183 Chr.9: 130595455-130595455 on Build GRCh38
rs201274970 Chr.9: 130595563-130595563 on Build GRCh38
rs769177757 Chr.9: 130595614-130595615 on Build GRCh38
rs768739903 Chr.9: 130595454-130595454 on Build GRCh38
rs750658354 Chr.9: 130595484-130595484 on Build GRCh38
rs999640536 Chr.9: 130595528-130595528 on Build GRCh38
rs755822691 Chr.9: 130595561-130595561 on Build GRCh38
rs761521159 Chr.9: 130595475-130595475 on Build GRCh38
rs903000398 Chr.9: 130595519-130595519 on Build GRCh38
rs368692366 Chr.9: 130595449-130595449 on Build GRCh38
rs748106600 Chr.9: 130595579-130595579 on Build GRCh38
rs768219076 Chr.9: 130595571-130595571 on Build GRCh38
rs748825354 Chr.9: 130595570-130595570 on Build GRCh38
rs771799360 Chr.9: 130595587-130595587 on Build GRCh38
rs200486785 Chr.9: 130595558-130595558 on Build GRCh38
rs753933525 Chr.9: 130595510-130595510 on Build GRCh38
rs749369285 Chr.9: 130595600-130595600 on Build GRCh38
rs1048515593 Chr.9: 130595529-130595529 on Build GRCh38
rs931489672 Chr.9: 130595505-130595505 on Build GRCh38
rs776766975 Chr.9: 130595606-130595606 on Build GRCh38
rs372290024 Chr.9: 130595498-130595498 on Build GRCh38
rs758746250 Chr.9: 130595542-130595542 on Build GRCh38
rs764467994 Chr.9: 130595638-130595638 on Build GRCh38
rs895473599 Chr.9: 130595565-130595565 on Build GRCh38
rs374300531 Chr.9: 130595442-130595442 on Build GRCh38
rs753195838 Chr.9: 130595541-130595541 on Build GRCh38
rs765737321 Chr.9: 130595533-130595533 on Build GRCh38
rs769238240 Chr.9: 130595433-130595433 on Build GRCh38
rs756351050 Chr.9: 130595454-130595457 on Build GRCh38
rs758909618 Chr.9: 130595635-130595635 on Build GRCh38
rs778179333 Chr.9: 130595557-130595557 on Build GRCh38
rs949927081 Chr.9: 130595431-130595431 on Build GRCh38
rs760102020 Chr.9: 130595575-130595575 on Build GRCh38
rs148816776 Chr.9: 130595504-130595504 on Build GRCh38
rs767344023 Chr.9: 130595479-130595479 on Build GRCh38
rs760334380 Chr.9: 130595599-130595599 on Build GRCh38
rs780034309 Chr.9: 130595460-130595460 on Build GRCh38
rs766597384 Chr.9: 130595600-130595600 on Build GRCh38
rs774499750 Chr.9: 130595574-130595574 on Build GRCh38
rs773088031 Chr.9: 130595598-130595598 on Build GRCh38
rs753194667 Chr.9: 130595630-130595630 on Build GRCh38
rs965986541 Chr.9: 130595625-130595625 on Build GRCh38
rs765286764 Chr.9: 130595611-130595611 on Build GRCh38

View Full Product Details