Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16049836-16050099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4 NM_001165945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2088612 c.937A>G p.N313D Chr.1: 16049885-16049885 on Build GRCh38
6873515 c.960G>A p.L320L Chr.1: 16049908-16049908 on Build GRCh38
6629941 c.962C>T p.A321V Chr.1: 16049910-16049910 on Build GRCh38
4512827 c.918C>T p.F306F Chr.1: 16049866-16049866 on Build GRCh38
4224655 c.430A>G p.N144D Chr.1: 16049885-16049885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553717185 Chr.1: 16049993-16049993 on Build GRCh38
rs549163396 Chr.1: 16049939-16049939 on Build GRCh38
rs962524610 Chr.1: 16050035-16050035 on Build GRCh38
rs145464420 Chr.1: 16050024-16050024 on Build GRCh38
rs776900398 Chr.1: 16049921-16049921 on Build GRCh38
rs12138156 Chr.1: 16050064-16050064 on Build GRCh38
rs111245746 Chr.1: 16049952-16049952 on Build GRCh38
rs1050769965 Chr.1: 16049887-16049887 on Build GRCh38
rs571586564 Chr.1: 16049962-16049962 on Build GRCh38
rs200650615 Chr.1: 16049925-16049925 on Build GRCh38
rs909824403 Chr.1: 16049998-16049998 on Build GRCh38
rs753346510 Chr.1: 16049957-16049957 on Build GRCh38
rs779581559 Chr.1: 16049893-16049893 on Build GRCh38
rs747309651 Chr.1: 16049911-16049911 on Build GRCh38
rs371225926 Chr.1: 16049941-16049941 on Build GRCh38
rs551691887 Chr.1: 16049959-16049959 on Build GRCh38
rs779593707 Chr.1: 16049884-16049885 on Build GRCh38
rs72651918 Chr.1: 16049999-16049999 on Build GRCh38
rs748649125 Chr.1: 16049903-16049903 on Build GRCh38
rs780933188 Chr.1: 16049876-16049876 on Build GRCh38
rs370023856 Chr.1: 16049890-16049890 on Build GRCh38
rs372394893 Chr.1: 16049924-16049924 on Build GRCh38
rs867576007 Chr.1: 16050023-16050023 on Build GRCh38
rs372714615 Chr.1: 16049954-16049954 on Build GRCh38
rs751411835 Chr.1: 16049943-16049943 on Build GRCh38
rs748661435 Chr.1: 16049955-16049956 on Build GRCh38
rs146865861 Chr.1: 16050026-16050026 on Build GRCh38
rs778139626 Chr.1: 16049906-16049906 on Build GRCh38
rs980424368 Chr.1: 16049975-16049975 on Build GRCh38
rs569455220 Chr.1: 16049950-16049950 on Build GRCh38
rs767415349 Chr.1: 16049951-16049951 on Build GRCh38
rs767468541 Chr.1: 16049967-16049967 on Build GRCh38
rs374011061 Chr.1: 16049963-16049963 on Build GRCh38
rs758937398 Chr.1: 16049956-16049957 on Build GRCh38
rs7368166 Chr.1: 16049963-16049963 on Build GRCh38
rs773010199 Chr.1: 16049968-16049968 on Build GRCh38
rs772579714 Chr.1: 16049904-16049904 on Build GRCh38
rs756972165 Chr.1: 16049859-16049859 on Build GRCh38
rs368835093 Chr.1: 16049892-16049892 on Build GRCh38
rs374950020 Chr.1: 16049961-16049961 on Build GRCh38
rs546971062 Chr.1: 16049856-16049856 on Build GRCh38
rs71003232 Chr.1: 16050023-16050024 on Build GRCh38
rs529399128 Chr.1: 16049929-16049929 on Build GRCh38
rs768840023 Chr.1: 16049964-16049964 on Build GRCh38
rs377215024 Chr.1: 16049858-16049858 on Build GRCh38
rs201204502 Chr.1: 16049917-16049917 on Build GRCh38
rs747502186 Chr.1: 16049960-16049960 on Build GRCh38

View Full Product Details