Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109914146-109914417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1
Entrez Gene ID
1435
Gene Name
colony stimulating factor 1
Gene Aliases
CSF-1, MCSF
UniGene
Hs.173894
Gene Chromosome Location
Chr.1: 109910506-109930994 on Build GRCh38
Transcripts
NM_000757.5 NM_172210.2 NM_172211.3 NM_172212.2 XM_017000369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636004 c.80C>T p.A27V Chr.1: 109914299-109914299 on Build GRCh38
1688247 c.131G>A p.G44E Chr.1: 109914350-109914350 on Build GRCh38
4467627 c.149C>T p.S50F Chr.1: 109914368-109914368 on Build GRCh38
7169510 c.75C>T p.L25L Chr.1: 109914294-109914294 on Build GRCh38
3788404 c.126G>A p.M42I Chr.1: 109914345-109914345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775348895 Chr.1: 109914385-109914385 on Build GRCh38
rs1012169952 Chr.1: 109914378-109914378 on Build GRCh38
rs574762856 Chr.1: 109914185-109914185 on Build GRCh38
rs747226112 Chr.1: 109914366-109914366 on Build GRCh38
rs372342555 Chr.1: 109914388-109914388 on Build GRCh38
rs762383396 Chr.1: 109914289-109914289 on Build GRCh38
rs758685697 Chr.1: 109914229-109914229 on Build GRCh38
rs552699078 Chr.1: 109914231-109914231 on Build GRCh38
rs200522461 Chr.1: 109914370-109914370 on Build GRCh38
rs540659231 Chr.1: 109914188-109914188 on Build GRCh38
rs774978352 Chr.1: 109914269-109914269 on Build GRCh38
rs765908652 Chr.1: 109914320-109914320 on Build GRCh38
rs749244779 Chr.1: 109914387-109914387 on Build GRCh38
rs894985163 Chr.1: 109914376-109914376 on Build GRCh38
rs371617668 Chr.1: 109914225-109914225 on Build GRCh38
rs532692920 Chr.1: 109914205-109914205 on Build GRCh38
rs758507202 Chr.1: 109914335-109914335 on Build GRCh38
rs202209718 Chr.1: 109914299-109914299 on Build GRCh38
rs550625120 Chr.1: 109914250-109914250 on Build GRCh38
rs748292286 Chr.1: 109914220-109914220 on Build GRCh38
rs200624006 Chr.1: 109914248-109914248 on Build GRCh38
rs998933506 Chr.1: 109914348-109914348 on Build GRCh38
rs755526140 Chr.1: 109914316-109914316 on Build GRCh38
rs754201417 Chr.1: 109914315-109914315 on Build GRCh38
rs766733065 Chr.1: 109914310-109914310 on Build GRCh38
rs71665078 Chr.1: 109914375-109914375 on Build GRCh38
rs745730488 Chr.1: 109914383-109914383 on Build GRCh38
rs903497730 Chr.1: 109914391-109914391 on Build GRCh38
rs767821994 Chr.1: 109914206-109914206 on Build GRCh38
rs2297706 Chr.1: 109914244-109914244 on Build GRCh38
rs191317982 Chr.1: 109914190-109914190 on Build GRCh38
rs34714942 Chr.1: 109914377-109914377 on Build GRCh38
rs1057496972 Chr.1: 109914199-109914199 on Build GRCh38
rs747220763 Chr.1: 109914243-109914243 on Build GRCh38
rs761402682 Chr.1: 109914308-109914308 on Build GRCh38
rs145602744 Chr.1: 109914294-109914294 on Build GRCh38
rs769581481 Chr.1: 109914384-109914384 on Build GRCh38
rs769783123 Chr.1: 109914268-109914268 on Build GRCh38
rs753257305 Chr.1: 109914326-109914326 on Build GRCh38
rs567581123 Chr.1: 109914365-109914365 on Build GRCh38
rs922111487 Chr.1: 109914170-109914170 on Build GRCh38
rs774005239 Chr.1: 109914304-109914304 on Build GRCh38
rs1013292274 Chr.1: 109914215-109914215 on Build GRCh38

View Full Product Details