Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 829263-829510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2B
Entrez Gene ID
283229
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
EFCAB4A
UniGene
Hs.660936
Gene Chromosome Location
Chr.11: 825558-831991 on Build GRCh38
Transcripts
NM_001286606.1 NM_173584.4 XM_011520011.1 XM_011520019.1 XM_017017581.1 XM_017017582.1 XM_017017583.1 XM_017017584.1 XM_017017585.1 XM_017017586.1 XM_017017587.1 XM_017017588.1 XM_017017589.1 XM_017017590.1 XM_017017591.1 XM_017017592.1 XM_017017593.1 XM_017017594.1 XM_017017595.1 XM_017017596.1 XM_017017597.1 XM_017017598.1 XM_017017599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422074 c.425T>C p.L142P Chr.11: 829507-829507 on Build GRCh38
5572036 c.286G>C p.V96L Chr.11: 829368-829368 on Build GRCh38
5462098 c.278-4G>A p.? Chr.11: 829356-829356 on Build GRCh38
3810549 c.328C>T p.P110S Chr.11: 829410-829410 on Build GRCh38
5147377 c.292G>A p.V98M Chr.11: 829374-829374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527935491 Chr.11: 829282-829282 on Build GRCh38
rs770145826 Chr.11: 829313-829313 on Build GRCh38
rs572460035 Chr.11: 829460-829460 on Build GRCh38
rs371971539 Chr.11: 829323-829323 on Build GRCh38
rs770585537 Chr.11: 829413-829415 on Build GRCh38
rs539907988 Chr.11: 829351-829351 on Build GRCh38
rs965923612 Chr.11: 829327-829327 on Build GRCh38
rs780259041 Chr.11: 829320-829320 on Build GRCh38
rs780852956 Chr.11: 829469-829470 on Build GRCh38
rs772686357 Chr.11: 829412-829412 on Build GRCh38
rs945588235 Chr.11: 829424-829424 on Build GRCh38
rs755007968 Chr.11: 829362-829362 on Build GRCh38
rs760789005 Chr.11: 829353-829353 on Build GRCh38
rs774770107 Chr.11: 829479-829481 on Build GRCh38
rs371372260 Chr.11: 829321-829321 on Build GRCh38
rs751094728 Chr.11: 829285-829285 on Build GRCh38
rs1051953440 Chr.11: 829435-829435 on Build GRCh38
rs376592119 Chr.11: 829474-829474 on Build GRCh38
rs778406267 Chr.11: 829475-829475 on Build GRCh38
rs868775289 Chr.11: 829359-829359 on Build GRCh38
rs557492056 Chr.11: 829425-829425 on Build GRCh38
rs761922699 Chr.11: 829344-829344 on Build GRCh38
rs771003508 Chr.11: 829482-829482 on Build GRCh38
rs752637387 Chr.11: 829374-829374 on Build GRCh38
rs376708795 Chr.11: 829415-829415 on Build GRCh38
rs1032721405 Chr.11: 829301-829301 on Build GRCh38
rs777419505 Chr.11: 829310-829310 on Build GRCh38
rs369732603 Chr.11: 829434-829434 on Build GRCh38
rs745330588 Chr.11: 829470-829472 on Build GRCh38
rs941453244 Chr.11: 829371-829371 on Build GRCh38
rs547963982 Chr.11: 829303-829303 on Build GRCh38
rs756848920 Chr.11: 829454-829454 on Build GRCh38
rs570965447 Chr.11: 829312-829312 on Build GRCh38
rs761036648 Chr.11: 829479-829479 on Build GRCh38
rs570060552 Chr.11: 829357-829357 on Build GRCh38
rs776795542 Chr.11: 829484-829484 on Build GRCh38
rs540832246 Chr.11: 829405-829405 on Build GRCh38
rs769304481 Chr.11: 829473-829481 on Build GRCh38
rs755351535 Chr.11: 829467-829467 on Build GRCh38
rs535466893 Chr.11: 829389-829389 on Build GRCh38
rs767216953 Chr.11: 829456-829456 on Build GRCh38
rs201767540 Chr.11: 829476-829476 on Build GRCh38
rs373571452 Chr.11: 829462-829462 on Build GRCh38
rs769152577 Chr.11: 829404-829404 on Build GRCh38
rs976375254 Chr.11: 829341-829341 on Build GRCh38
rs762775852 Chr.11: 829448-829448 on Build GRCh38
rs751027318 Chr.11: 829450-829450 on Build GRCh38
rs750101100 Chr.11: 829464-829464 on Build GRCh38
rs765049710 Chr.11: 829443-829443 on Build GRCh38
rs748631052 Chr.11: 829408-829408 on Build GRCh38
rs909958914 Chr.11: 829361-829361 on Build GRCh38
rs773138241 Chr.11: 829413-829413 on Build GRCh38
rs555677202 Chr.11: 829392-829392 on Build GRCh38
rs200792273 Chr.11: 829393-829393 on Build GRCh38
rs749657369 Chr.11: 829394-829394 on Build GRCh38
rs769281432 Chr.11: 829322-829322 on Build GRCh38
rs752584365 Chr.11: 829447-829447 on Build GRCh38
rs765101154 Chr.11: 829370-829370 on Build GRCh38
rs748588923 Chr.11: 829472-829472 on Build GRCh38
rs781272807 Chr.11: 829372-829372 on Build GRCh38
rs372110906 Chr.11: 829379-829379 on Build GRCh38
rs757886314 Chr.11: 829377-829377 on Build GRCh38
rs149293789 Chr.11: 829356-829356 on Build GRCh38
rs772046526 Chr.11: 829350-829350 on Build GRCh38
rs764130209 Chr.11: 829449-829449 on Build GRCh38

View Full Product Details