Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15347123-15347396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGF
Entrez Gene ID
2277
Gene Name
c-fos induced growth factor
Gene Aliases
VEGF-D, VEGFD
UniGene
Hs.11392
Gene Chromosome Location
Chr.X: 15345591-15384413 on Build GRCh38
Transcripts
NM_004469.4 NR_037859.2

Gene Symbol
PIR-FIGF
Entrez Gene ID
100532742
Gene Name
PIR-FIGF readthrough
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15345591-15491309 on Build GRCh38
Transcripts
NM_004469.4 NR_037859.2

SNP Detail

SNP ID Chromosome Location
rs192921923 Chr.X: 15347247-15347247 on Build GRCh38
rs368069978 Chr.X: 15347268-15347268 on Build GRCh38
rs781296967 Chr.X: 15347205-15347205 on Build GRCh38
rs774553099 Chr.X: 15347226-15347226 on Build GRCh38
rs779966228 Chr.X: 15347192-15347192 on Build GRCh38
rs370603238 Chr.X: 15347300-15347300 on Build GRCh38
rs780465926 Chr.X: 15347180-15347180 on Build GRCh38
rs772183457 Chr.X: 15347237-15347237 on Build GRCh38
rs750707563 Chr.X: 15347152-15347152 on Build GRCh38
rs765960915 Chr.X: 15347306-15347306 on Build GRCh38
rs757857479 Chr.X: 15347305-15347305 on Build GRCh38
rs757971531 Chr.X: 15347190-15347190 on Build GRCh38
rs748142888 Chr.X: 15347320-15347320 on Build GRCh38
rs777155942 Chr.X: 15347276-15347276 on Build GRCh38
rs756676683 Chr.X: 15347156-15347156 on Build GRCh38
rs780418465 Chr.X: 15347351-15347351 on Build GRCh38
rs748056784 Chr.X: 15347215-15347215 on Build GRCh38
rs750123121 Chr.X: 15347187-15347187 on Build GRCh38
rs775717720 Chr.X: 15347246-15347246 on Build GRCh38
rs746783235 Chr.X: 15347204-15347204 on Build GRCh38
rs147950971 Chr.X: 15347269-15347269 on Build GRCh38
rs141907720 Chr.X: 15347299-15347299 on Build GRCh38
rs762279003 Chr.X: 15347295-15347295 on Build GRCh38
rs756109504 Chr.X: 15347352-15347352 on Build GRCh38
rs771172462 Chr.X: 15347218-15347218 on Build GRCh38
rs146086561 Chr.X: 15347316-15347316 on Build GRCh38
rs751171329 Chr.X: 15347307-15347307 on Build GRCh38
rs754847710 Chr.X: 15347314-15347314 on Build GRCh38

View Full Product Details