Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121899805-121900074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATE1
Entrez Gene ID
11101
Gene Name
arginyltransferase 1
Gene Aliases
-
UniGene
Hs.632080
Gene Chromosome Location
Chr.10: 121740421-121928801 on Build GRCh38
Transcripts
NM_001001976.2 NM_001288734.1 NM_001288735.1 NM_001288736.1 NM_007041.3 NR_110006.1 XM_005269458.2 XM_005269459.3 XM_017015597.1 XM_017015598.1 XM_017015599.1 XM_017015600.1 XM_017015601.1 XM_017015602.1 XM_017015603.1 XM_017015604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6319979 c.939C>A p.S313R Chr.10: 121899869-121899869 on Build GRCh38
6606500 c.833C>A p.S278Y Chr.10: 121899975-121899975 on Build GRCh38
5753420 c.846G>A p.S282S Chr.10: 121899962-121899962 on Build GRCh38
5503669 c.885T>A p.Y295* Chr.10: 121899923-121899923 on Build GRCh38
279024 c.856G>T p.A286S Chr.10: 121899952-121899952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150104028 Chr.10: 121899874-121899874 on Build GRCh38
rs762200087 Chr.10: 121899910-121899910 on Build GRCh38
rs766867330 Chr.10: 121899928-121899928 on Build GRCh38
rs779889930 Chr.10: 121899989-121899989 on Build GRCh38
rs763630233 Chr.10: 121899914-121899915 on Build GRCh38
rs768017074 Chr.10: 121899918-121899918 on Build GRCh38
rs567669216 Chr.10: 121899963-121899963 on Build GRCh38
rs754103610 Chr.10: 121899931-121899931 on Build GRCh38
rs370614627 Chr.10: 121899990-121899990 on Build GRCh38
rs766849302 Chr.10: 121900040-121900040 on Build GRCh38
rs768399563 Chr.10: 121899996-121899996 on Build GRCh38
rs1056846302 Chr.10: 121899884-121899884 on Build GRCh38
rs745816449 Chr.10: 121899967-121899967 on Build GRCh38
rs905500969 Chr.10: 121899875-121899875 on Build GRCh38
rs770942976 Chr.10: 121899858-121899858 on Build GRCh38
rs773947063 Chr.10: 121899903-121899903 on Build GRCh38
rs202159245 Chr.10: 121900006-121900006 on Build GRCh38
rs868503526 Chr.10: 121899853-121899853 on Build GRCh38
rs751270541 Chr.10: 121899920-121899921 on Build GRCh38
rs758431848 Chr.10: 121899962-121899962 on Build GRCh38
rs778828743 Chr.10: 121899947-121899947 on Build GRCh38
rs1028593693 Chr.10: 121900009-121900009 on Build GRCh38
rs373331175 Chr.10: 121899924-121899924 on Build GRCh38
rs750922363 Chr.10: 121899919-121899919 on Build GRCh38
rs374263772 Chr.10: 121899849-121899849 on Build GRCh38
rs370475140 Chr.10: 121899878-121899878 on Build GRCh38
rs201217964 Chr.10: 121900043-121900043 on Build GRCh38
rs761325742 Chr.10: 121900031-121900031 on Build GRCh38
rs200676873 Chr.10: 121899960-121899960 on Build GRCh38
rs755888288 Chr.10: 121899976-121899976 on Build GRCh38
rs200592546 Chr.10: 121900000-121900000 on Build GRCh38
rs887492651 Chr.10: 121899939-121899939 on Build GRCh38
rs755055072 Chr.10: 121899839-121899839 on Build GRCh38
rs746916204 Chr.10: 121899857-121899857 on Build GRCh38
rs746597136 Chr.10: 121899925-121899926 on Build GRCh38
rs773800669 Chr.10: 121900017-121900017 on Build GRCh38
rs769579445 Chr.10: 121899876-121899876 on Build GRCh38
rs756965864 Chr.10: 121900008-121900009 on Build GRCh38
rs758011917 Chr.10: 121899882-121899883 on Build GRCh38
rs779111565 Chr.10: 121899848-121899848 on Build GRCh38
rs1044320677 Chr.10: 121899852-121899852 on Build GRCh38
rs752526283 Chr.10: 121899844-121899850 on Build GRCh38
rs374428502 Chr.10: 121899997-121899997 on Build GRCh38
rs755322923 Chr.10: 121899936-121899936 on Build GRCh38

View Full Product Details