Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43494407-43494676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
UniGene
Hs.661641
Gene Chromosome Location
Chr.5: 43486699-43515433 on Build GRCh38
Transcripts
NM_198566.3 XM_011514038.2 XM_017009445.1 XM_017009446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855370 c.1179T>C p.N393N Chr.5: 43494575-43494575 on Build GRCh38
6470398 c.1228A>T p.I410F Chr.5: 43494526-43494526 on Build GRCh38
6470397 c.1229T>A p.I410N Chr.5: 43494525-43494525 on Build GRCh38
7310202 c.1176A>T p.V392V Chr.5: 43494578-43494578 on Build GRCh38
3342714 c.1172C>T p.S391L Chr.5: 43494582-43494582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760651416 Chr.5: 43494490-43494490 on Build GRCh38
rs377319549 Chr.5: 43494629-43494629 on Build GRCh38
rs746322083 Chr.5: 43494520-43494520 on Build GRCh38
rs759536999 Chr.5: 43494498-43494498 on Build GRCh38
rs775607313 Chr.5: 43494609-43494609 on Build GRCh38
rs974863171 Chr.5: 43494449-43494449 on Build GRCh38
rs964694199 Chr.5: 43494434-43494434 on Build GRCh38
rs371448584 Chr.5: 43494487-43494487 on Build GRCh38
rs149386441 Chr.5: 43494491-43494491 on Build GRCh38
rs751650632 Chr.5: 43494610-43494610 on Build GRCh38
rs564658256 Chr.5: 43494554-43494554 on Build GRCh38
rs781547965 Chr.5: 43494519-43494519 on Build GRCh38
rs1023917002 Chr.5: 43494595-43494595 on Build GRCh38
rs771686166 Chr.5: 43494538-43494538 on Build GRCh38
rs1030117952 Chr.5: 43494543-43494543 on Build GRCh38
rs765262832 Chr.5: 43494598-43494598 on Build GRCh38
rs754462830 Chr.5: 43494534-43494534 on Build GRCh38
rs767867885 Chr.5: 43494635-43494635 on Build GRCh38
rs771880278 Chr.5: 43494484-43494484 on Build GRCh38
rs375537856 Chr.5: 43494472-43494472 on Build GRCh38
rs757721389 Chr.5: 43494555-43494555 on Build GRCh38
rs757807463 Chr.5: 43494470-43494470 on Build GRCh38
rs773045303 Chr.5: 43494544-43494544 on Build GRCh38
rs776835412 Chr.5: 43494497-43494497 on Build GRCh38
rs768423993 Chr.5: 43494473-43494473 on Build GRCh38
rs1000022642 Chr.5: 43494578-43494578 on Build GRCh38
rs113706450 Chr.5: 43494545-43494545 on Build GRCh38
rs763315590 Chr.5: 43494618-43494618 on Build GRCh38
rs747549953 Chr.5: 43494465-43494465 on Build GRCh38
rs764705867 Chr.5: 43494507-43494508 on Build GRCh38
rs139034792 Chr.5: 43494516-43494516 on Build GRCh38
rs936930061 Chr.5: 43494475-43494475 on Build GRCh38
rs147140419 Chr.5: 43494536-43494536 on Build GRCh38
rs995987375 Chr.5: 43494542-43494542 on Build GRCh38
rs1044161038 Chr.5: 43494515-43494515 on Build GRCh38
rs765300404 Chr.5: 43494510-43494510 on Build GRCh38
rs370679151 Chr.5: 43494461-43494461 on Build GRCh38
rs1031062591 Chr.5: 43494579-43494579 on Build GRCh38
rs375174167 Chr.5: 43494517-43494517 on Build GRCh38
rs758845266 Chr.5: 43494459-43494459 on Build GRCh38
rs537002772 Chr.5: 43494479-43494479 on Build GRCh38
rs769910457 Chr.5: 43494463-43494464 on Build GRCh38
rs989485922 Chr.5: 43494584-43494584 on Build GRCh38
rs371853399 Chr.5: 43494555-43494555 on Build GRCh38
rs746798968 Chr.5: 43494546-43494546 on Build GRCh38
rs764399181 Chr.5: 43494628-43494628 on Build GRCh38
rs779759619 Chr.5: 43494471-43494471 on Build GRCh38
rs938423788 Chr.5: 43494458-43494458 on Build GRCh38
rs750918336 Chr.5: 43494648-43494648 on Build GRCh38
rs200027263 Chr.5: 43494582-43494582 on Build GRCh38
rs143374727 Chr.5: 43494535-43494535 on Build GRCh38
rs369798820 Chr.5: 43494518-43494518 on Build GRCh38
rs368627268 Chr.5: 43494474-43494474 on Build GRCh38
rs754340959 Chr.5: 43494522-43494525 on Build GRCh38

View Full Product Details