Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76505281-76505510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX2
Entrez Gene ID
113878
Gene Name
deltex E3 ubiquitin ligase 2
Gene Aliases
RNF58
UniGene
Hs.187058
Gene Chromosome Location
Chr.7: 76461655-76505995 on Build GRCh38
Transcripts
NM_001102594.1 NM_001102595.1 NM_001102596.1 NM_020892.2 XM_005250131.1 XM_005250132.1 XM_011515772.1 XM_011515773.2 XM_011515774.1 XM_011515775.1 XM_017011723.1 XM_017011724.1 XM_017011725.1 XM_017011726.1 XM_017011727.1 XM_017011728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291441 c.1676G>T p.R559L Chr.7: 76505408-76505408 on Build GRCh38
4471145 c.1701C>T p.S567S Chr.7: 76505433-76505433 on Build GRCh38
3698547 c.1680C>T p.L560L Chr.7: 76505412-76505412 on Build GRCh38
1452122 c.1725C>T p.T575T Chr.7: 76505457-76505457 on Build GRCh38
5873098 c.1684T>C p.F562L Chr.7: 76505416-76505416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037542705 Chr.7: 76505343-76505343 on Build GRCh38
rs910358488 Chr.7: 76505304-76505304 on Build GRCh38
rs772565087 Chr.7: 76505427-76505427 on Build GRCh38
rs566691920 Chr.7: 76505300-76505300 on Build GRCh38
rs139355667 Chr.7: 76505451-76505451 on Build GRCh38
rs759881389 Chr.7: 76505331-76505331 on Build GRCh38
rs187546018 Chr.7: 76505450-76505450 on Build GRCh38
rs998516653 Chr.7: 76505484-76505484 on Build GRCh38
rs765387474 Chr.7: 76505320-76505320 on Build GRCh38
rs750652106 Chr.7: 76505444-76505444 on Build GRCh38
rs374855456 Chr.7: 76505359-76505359 on Build GRCh38
rs141181111 Chr.7: 76505408-76505408 on Build GRCh38
rs778500050 Chr.7: 76505470-76505470 on Build GRCh38
rs781172309 Chr.7: 76505368-76505368 on Build GRCh38
rs776131330 Chr.7: 76505335-76505335 on Build GRCh38
rs73363181 Chr.7: 76505412-76505412 on Build GRCh38
rs748045937 Chr.7: 76505372-76505372 on Build GRCh38
rs752437258 Chr.7: 76505455-76505455 on Build GRCh38
rs376786877 Chr.7: 76505469-76505469 on Build GRCh38
rs567492749 Chr.7: 76505357-76505357 on Build GRCh38
rs929315264 Chr.7: 76505358-76505358 on Build GRCh38
rs774607429 Chr.7: 76505329-76505329 on Build GRCh38
rs748763928 Chr.7: 76505458-76505458 on Build GRCh38
rs985863930 Chr.7: 76505303-76505303 on Build GRCh38
rs751323273 Chr.7: 76505365-76505365 on Build GRCh38
rs764614517 Chr.7: 76505433-76505433 on Build GRCh38
rs545347146 Chr.7: 76505441-76505441 on Build GRCh38
rs768155478 Chr.7: 76505334-76505334 on Build GRCh38
rs368665521 Chr.7: 76505430-76505430 on Build GRCh38
rs373052973 Chr.7: 76505436-76505436 on Build GRCh38
rs761242077 Chr.7: 76505432-76505432 on Build GRCh38
rs755665034 Chr.7: 76505380-76505380 on Build GRCh38
rs773999130 Chr.7: 76505422-76505422 on Build GRCh38
rs765433775 Chr.7: 76505442-76505442 on Build GRCh38
rs770491166 Chr.7: 76505421-76505421 on Build GRCh38
rs756074116 Chr.7: 76505457-76505457 on Build GRCh38
rs201931701 Chr.7: 76505340-76505340 on Build GRCh38
rs754752737 Chr.7: 76505367-76505367 on Build GRCh38
rs746295240 Chr.7: 76505425-76505425 on Build GRCh38
rs764238019 Chr.7: 76505346-76505346 on Build GRCh38
rs73140090 Chr.7: 76505356-76505356 on Build GRCh38
rs753243070 Chr.7: 76505355-76505357 on Build GRCh38
rs745488629 Chr.7: 76505471-76505471 on Build GRCh38

View Full Product Details