Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28590045-28590296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGF29
Entrez Gene ID
112869
Gene Name
SAGA complex associated factor 29
Gene Aliases
CCDC101, STAF36, TDRD29
UniGene
Hs.655476
Gene Chromosome Location
Chr.16: 28553915-28591790 on Build GRCh38
Transcripts
NM_138414.2 XM_017022894.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3109786 c.372G>A p.Q124Q Chr.16: 28590178-28590178 on Build GRCh38
1742935 c.391C>T p.P131S Chr.16: 28590197-28590197 on Build GRCh38
7313283 c.358A>T p.M120L Chr.16: 28590164-28590164 on Build GRCh38
140088 c.362C>T p.T121I Chr.16: 28590168-28590168 on Build GRCh38
969485 c.304G>A p.G102S Chr.16: 28590110-28590110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748462252 Chr.16: 28590257-28590257 on Build GRCh38
rs367762239 Chr.16: 28590252-28590252 on Build GRCh38
rs763714883 Chr.16: 28590196-28590196 on Build GRCh38
rs574507193 Chr.16: 28590169-28590169 on Build GRCh38
rs181679812 Chr.16: 28590069-28590069 on Build GRCh38
rs774745658 Chr.16: 28590110-28590110 on Build GRCh38
rs779712141 Chr.16: 28590088-28590089 on Build GRCh38
rs781511958 Chr.16: 28590070-28590070 on Build GRCh38
rs758275357 Chr.16: 28590243-28590243 on Build GRCh38
rs989046905 Chr.16: 28590272-28590272 on Build GRCh38
rs769905203 Chr.16: 28590266-28590266 on Build GRCh38
rs925691702 Chr.16: 28590249-28590249 on Build GRCh38
rs577053958 Chr.16: 28590242-28590242 on Build GRCh38
rs2291939 Chr.16: 28590208-28590208 on Build GRCh38
rs530098702 Chr.16: 28590106-28590106 on Build GRCh38
rs955921451 Chr.16: 28590269-28590269 on Build GRCh38
rs533131207 Chr.16: 28590137-28590137 on Build GRCh38
rs767827594 Chr.16: 28590130-28590130 on Build GRCh38
rs769931603 Chr.16: 28590089-28590089 on Build GRCh38
rs372062198 Chr.16: 28590211-28590211 on Build GRCh38
rs748254585 Chr.16: 28590074-28590074 on Build GRCh38
rs773152836 Chr.16: 28590090-28590090 on Build GRCh38
rs1004481110 Chr.16: 28590142-28590142 on Build GRCh38
rs763519681 Chr.16: 28590097-28590097 on Build GRCh38
rs955120046 Chr.16: 28590086-28590086 on Build GRCh38
rs759915589 Chr.16: 28590127-28590127 on Build GRCh38
rs150312488 Chr.16: 28590109-28590109 on Build GRCh38
rs543411215 Chr.16: 28590184-28590184 on Build GRCh38

View Full Product Details