Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39835827-39836088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTHD1
Entrez Gene ID
150350
Gene Name
ENTH domain containing 1
Gene Aliases
CACNA1I, dJ370M22.3
UniGene
Hs.474869
Gene Chromosome Location
Chr.22: 39743044-39893884 on Build GRCh38
Transcripts
NM_152512.3 XM_006724149.3 XM_006724150.3 XM_006724151.1 XM_006724153.3 XM_011529927.2 XM_011529928.2 XM_011529931.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478985 c.638T>A p.V213D Chr.22: 39835913-39835913 on Build GRCh38
5465489 c.653A>C p.E218A Chr.22: 39835898-39835898 on Build GRCh38
1416378 c.669G>T p.Q223H Chr.22: 39835882-39835882 on Build GRCh38
3730412 c.647C>T p.P216L Chr.22: 39835904-39835904 on Build GRCh38
5530493 c.634G>A p.D212N Chr.22: 39835917-39835917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768501802 Chr.22: 39835917-39835917 on Build GRCh38
rs749796589 Chr.22: 39836003-39836003 on Build GRCh38
rs143255336 Chr.22: 39835911-39835911 on Build GRCh38
rs573700877 Chr.22: 39835890-39835890 on Build GRCh38
rs754623336 Chr.22: 39836003-39836007 on Build GRCh38
rs778492802 Chr.22: 39835991-39835991 on Build GRCh38
rs763080060 Chr.22: 39835938-39835938 on Build GRCh38
rs753439850 Chr.22: 39835992-39835992 on Build GRCh38
rs751784997 Chr.22: 39835954-39835954 on Build GRCh38
rs745413495 Chr.22: 39835876-39835876 on Build GRCh38
rs114100360 Chr.22: 39835936-39835936 on Build GRCh38
rs999326158 Chr.22: 39835975-39835975 on Build GRCh38
rs766390023 Chr.22: 39835944-39835944 on Build GRCh38
rs190190311 Chr.22: 39835981-39835981 on Build GRCh38
rs766107611 Chr.22: 39835977-39835977 on Build GRCh38
rs755115935 Chr.22: 39835968-39835968 on Build GRCh38
rs1038706344 Chr.22: 39835869-39835869 on Build GRCh38
rs756473600 Chr.22: 39835990-39835990 on Build GRCh38
rs755963500 Chr.22: 39835958-39835958 on Build GRCh38
rs1012156932 Chr.22: 39836046-39836046 on Build GRCh38
rs778403003 Chr.22: 39835871-39835871 on Build GRCh38
rs933111610 Chr.22: 39835889-39835889 on Build GRCh38
rs770151688 Chr.22: 39835868-39835868 on Build GRCh38
rs900161867 Chr.22: 39835885-39835885 on Build GRCh38
rs1038599176 Chr.22: 39836045-39836045 on Build GRCh38
rs534656585 Chr.22: 39835920-39835920 on Build GRCh38
rs760345892 Chr.22: 39835914-39835914 on Build GRCh38

View Full Product Details