Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101013675-101013902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH5
Entrez Gene ID
51611
Gene Name
diphthamide biosynthesis 5
Gene Aliases
AD-018, CGI-30, HSPC143, NPD015
UniGene
Hs.440776
Gene Chromosome Location
Chr.1: 100989623-101025806 on Build GRCh38
Transcripts
NM_001077394.1 NM_001077395.1 NM_015958.2 XM_005270938.1 XM_005270939.4 XM_005270941.3 XM_006710683.3 XM_011541569.1 XM_011541570.2 XM_017001460.1 XM_017001461.1 XM_017001462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6254849 c.293C>A p.A98E Chr.1: 101013786-101013786 on Build GRCh38
4415280 c.351A>T p.V117V Chr.1: 101013728-101013728 on Build GRCh38
3470332 c.354C>T p.G118G Chr.1: 101013725-101013725 on Build GRCh38
3671316 c.303G>T p.L101L Chr.1: 101013776-101013776 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776285372 Chr.1: 101013763-101013764 on Build GRCh38
rs778073486 Chr.1: 101013742-101013742 on Build GRCh38
rs756590567 Chr.1: 101013865-101013865 on Build GRCh38
rs368593058 Chr.1: 101013750-101013750 on Build GRCh38
rs761436132 Chr.1: 101013787-101013787 on Build GRCh38
rs766283412 Chr.1: 101013809-101013809 on Build GRCh38
rs372929823 Chr.1: 101013764-101013764 on Build GRCh38
rs745909194 Chr.1: 101013864-101013864 on Build GRCh38
rs753581555 Chr.1: 101013818-101013818 on Build GRCh38
rs375316542 Chr.1: 101013703-101013703 on Build GRCh38
rs369815138 Chr.1: 101013806-101013806 on Build GRCh38
rs1056891868 Chr.1: 101013756-101013756 on Build GRCh38
rs375522712 Chr.1: 101013796-101013796 on Build GRCh38
rs761351998 Chr.1: 101013860-101013861 on Build GRCh38
rs751879650 Chr.1: 101013840-101013840 on Build GRCh38
rs747229142 Chr.1: 101013748-101013748 on Build GRCh38
rs775736408 Chr.1: 101013769-101013769 on Build GRCh38
rs769006133 Chr.1: 101013780-101013780 on Build GRCh38
rs766119012 Chr.1: 101013730-101013730 on Build GRCh38
rs780871862 Chr.1: 101013753-101013753 on Build GRCh38
rs902987444 Chr.1: 101013747-101013747 on Build GRCh38
rs901597654 Chr.1: 101013823-101013823 on Build GRCh38
rs1016306230 Chr.1: 101013844-101013844 on Build GRCh38
rs772739169 Chr.1: 101013792-101013792 on Build GRCh38
rs34311144 Chr.1: 101013713-101013714 on Build GRCh38
rs746398589 Chr.1: 101013754-101013754 on Build GRCh38
rs765110401 Chr.1: 101013829-101013829 on Build GRCh38
rs753621373 Chr.1: 101013732-101013732 on Build GRCh38
rs757518300 Chr.1: 101013861-101013861 on Build GRCh38
rs75419073 Chr.1: 101013728-101013728 on Build GRCh38
rs781158743 Chr.1: 101013863-101013863 on Build GRCh38
rs758710149 Chr.1: 101013739-101013739 on Build GRCh38
rs1049040637 Chr.1: 101013766-101013766 on Build GRCh38
rs566025190 Chr.1: 101013848-101013848 on Build GRCh38
rs754884736 Chr.1: 101013824-101013824 on Build GRCh38
rs774051094 Chr.1: 101013785-101013785 on Build GRCh38
rs767058878 Chr.1: 101013788-101013788 on Build GRCh38
rs749593698 Chr.1: 101013773-101013773 on Build GRCh38

View Full Product Details