Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 108348454-108348697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL17
Entrez Gene ID
64839
Gene Name
F-box and leucine rich repeat protein 17
Gene Aliases
FBXO13, Fbl17, Fbx13
UniGene
Hs.657225
Gene Chromosome Location
Chr.5: 107859033-108382098 on Build GRCh38
Transcripts
NM_001163315.2 XM_005272048.4 XM_005272050.4 XM_011543574.2 XM_011543575.2 XM_011543576.2 XM_011543577.2 XM_011543578.2 XM_011543579.2 XM_011543580.2 XM_017009729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5939358 c.1408C>T p.H470Y Chr.5: 108348497-108348497 on Build GRCh38
4868779 c.199G>T p.E67* Chr.5: 108348512-108348512 on Build GRCh38
3409661 c.220G>A p.G74S Chr.5: 108348491-108348491 on Build GRCh38
4343870 c.217T>C p.F73L Chr.5: 108348494-108348494 on Build GRCh38
5939360 c.214C>T p.H72Y Chr.5: 108348497-108348497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047984756 Chr.5: 108348638-108348638 on Build GRCh38
rs140473817 Chr.5: 108348530-108348530 on Build GRCh38
rs566422588 Chr.5: 108348640-108348640 on Build GRCh38
rs910617086 Chr.5: 108348594-108348595 on Build GRCh38
rs750655133 Chr.5: 108348501-108348501 on Build GRCh38
rs111631050 Chr.5: 108348568-108348568 on Build GRCh38
rs370257212 Chr.5: 108348477-108348477 on Build GRCh38
rs199665067 Chr.5: 108348566-108348566 on Build GRCh38
rs755067120 Chr.5: 108348556-108348556 on Build GRCh38
rs764025263 Chr.5: 108348499-108348499 on Build GRCh38
rs567547695 Chr.5: 108348603-108348603 on Build GRCh38
rs749397664 Chr.5: 108348524-108348524 on Build GRCh38
rs772269833 Chr.5: 108348654-108348654 on Build GRCh38
rs943487071 Chr.5: 108348597-108348597 on Build GRCh38
rs995676056 Chr.5: 108348590-108348590 on Build GRCh38
rs1051772958 Chr.5: 108348585-108348585 on Build GRCh38
rs17161259 Chr.5: 108348558-108348558 on Build GRCh38
rs772403151 Chr.5: 108348559-108348559 on Build GRCh38
rs80125529 Chr.5: 108348612-108348612 on Build GRCh38
rs183842521 Chr.5: 108348522-108348522 on Build GRCh38
rs1862195 Chr.5: 108348623-108348623 on Build GRCh38
rs550929689 Chr.5: 108348589-108348589 on Build GRCh38
rs980819489 Chr.5: 108348631-108348631 on Build GRCh38
rs1026710238 Chr.5: 108348593-108348593 on Build GRCh38
rs779384844 Chr.5: 108348557-108348557 on Build GRCh38
rs779960651 Chr.5: 108348523-108348523 on Build GRCh38
rs775948720 Chr.5: 108348574-108348574 on Build GRCh38
rs951218594 Chr.5: 108348601-108348601 on Build GRCh38
rs763058660 Chr.5: 108348561-108348561 on Build GRCh38
rs770620044 Chr.5: 108348569-108348569 on Build GRCh38
rs200990649 Chr.5: 108348492-108348492 on Build GRCh38
rs147193389 Chr.5: 108348519-108348519 on Build GRCh38
rs890706269 Chr.5: 108348543-108348543 on Build GRCh38
rs752740515 Chr.5: 108348491-108348491 on Build GRCh38

View Full Product Details