Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69180979-69181220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4B
Entrez Gene ID
23150
Gene Name
FERM domain containing 4B
Gene Aliases
6030440G05Rik, GRSP1
UniGene
Hs.371681
Gene Chromosome Location
Chr.3: 69168782-69541340 on Build GRCh38
Transcripts
NM_015123.2 XM_005264722.1 XM_005264723.1 XM_017005989.1 XM_017005990.1 XM_017005991.1 XM_017005992.1 XM_017005993.1 XM_017005994.1 XM_017005995.1 XM_017005996.1 XM_017005997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6666967 c.2590C>T p.R864W Chr.3: 69181160-69181160 on Build GRCh38
7143783 c.2380G>C p.E794Q Chr.3: 69181208-69181208 on Build GRCh38
1729344 c.2467A>T p.R823W Chr.3: 69181121-69181121 on Build GRCh38
6006976 c.2599C>T p.H867Y Chr.3: 69181151-69181151 on Build GRCh38
4483982 c.2595C>T p.S865S Chr.3: 69180993-69180993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970718105 Chr.3: 69181009-69181009 on Build GRCh38
rs759229825 Chr.3: 69181062-69181062 on Build GRCh38
rs768102366 Chr.3: 69181170-69181170 on Build GRCh38
rs754064141 Chr.3: 69181180-69181180 on Build GRCh38
rs924344210 Chr.3: 69181153-69181153 on Build GRCh38
rs755167962 Chr.3: 69181196-69181196 on Build GRCh38
rs770786696 Chr.3: 69181141-69181141 on Build GRCh38
rs533947293 Chr.3: 69181051-69181051 on Build GRCh38
rs762417131 Chr.3: 69181166-69181166 on Build GRCh38
rs776150142 Chr.3: 69181144-69181144 on Build GRCh38
rs1016741477 Chr.3: 69181066-69181066 on Build GRCh38
rs61737530 Chr.3: 69181011-69181011 on Build GRCh38
rs771827489 Chr.3: 69181131-69181131 on Build GRCh38
rs764522166 Chr.3: 69181077-69181077 on Build GRCh38
rs747861249 Chr.3: 69181036-69181036 on Build GRCh38
rs1001263500 Chr.3: 69181013-69181013 on Build GRCh38
rs753127481 Chr.3: 69181070-69181070 on Build GRCh38
rs769852415 Chr.3: 69181018-69181018 on Build GRCh38
rs374219123 Chr.3: 69181158-69181158 on Build GRCh38
rs977148936 Chr.3: 69181121-69181121 on Build GRCh38
rs751918469 Chr.3: 69181162-69181162 on Build GRCh38
rs762664931 Chr.3: 69181160-69181160 on Build GRCh38
rs765927903 Chr.3: 69181059-69181059 on Build GRCh38
rs746153581 Chr.3: 69181016-69181016 on Build GRCh38
rs777328436 Chr.3: 69181147-69181147 on Build GRCh38
rs771875204 Chr.3: 69181047-69181047 on Build GRCh38
rs756184115 Chr.3: 69181175-69181175 on Build GRCh38
rs764755686 Chr.3: 69181068-69181068 on Build GRCh38
rs757455228 Chr.3: 69181012-69181012 on Build GRCh38
rs746960161 Chr.3: 69181138-69181138 on Build GRCh38
rs780173818 Chr.3: 69181176-69181176 on Build GRCh38
rs780094507 Chr.3: 69181122-69181122 on Build GRCh38
rs776060332 Chr.3: 69181061-69181061 on Build GRCh38
rs769325621 Chr.3: 69181151-69181151 on Build GRCh38
rs778094939 Chr.3: 69181004-69181004 on Build GRCh38
rs376555402 Chr.3: 69181052-69181052 on Build GRCh38
rs758920846 Chr.3: 69181150-69181150 on Build GRCh38
rs202195281 Chr.3: 69181033-69181033 on Build GRCh38
rs781417922 Chr.3: 69181015-69181015 on Build GRCh38
rs781438145 Chr.3: 69181094-69181094 on Build GRCh38
rs769309837 Chr.3: 69181102-69181102 on Build GRCh38
rs569053028 Chr.3: 69181101-69181101 on Build GRCh38
rs750542052 Chr.3: 69181172-69181172 on Build GRCh38
rs753553130 Chr.3: 69181001-69181001 on Build GRCh38
rs540273782 Chr.3: 69181003-69181003 on Build GRCh38
rs764612126 Chr.3: 69181161-69181161 on Build GRCh38
rs749491850 Chr.3: 69181129-69181129 on Build GRCh38
rs552752466 Chr.3: 69181080-69181080 on Build GRCh38
rs370212737 Chr.3: 69181002-69181002 on Build GRCh38
rs192984298 Chr.3: 69181073-69181073 on Build GRCh38
rs747478566 Chr.3: 69181079-69181079 on Build GRCh38
rs1010403497 Chr.3: 69181091-69181091 on Build GRCh38

View Full Product Details