Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50320207-50320434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
UniGene
Hs.437894
Gene Chromosome Location
Chr.16: 50317563-50368969 on Build GRCh38
Transcripts
NM_001173984.2 NM_013263.4 XM_011523046.2 XM_011523047.2 XM_011523048.2 XM_011523049.2 XM_011523050.2 XM_017023179.1 XM_017023180.1 XM_017023181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5504203 c.1677C>A p.A559A Chr.16: 50320327-50320327 on Build GRCh38
7146956 c.1652G>C p.R551T Chr.16: 50320352-50320352 on Build GRCh38
3510034 c.1709C>T p.P570L Chr.16: 50320295-50320295 on Build GRCh38
1378187 c.1656G>A p.L552L Chr.16: 50320348-50320348 on Build GRCh38
1176754 c.1652G>A p.R551K Chr.16: 50320352-50320352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772936465 Chr.16: 50320295-50320295 on Build GRCh38
rs770797884 Chr.16: 50320301-50320301 on Build GRCh38
rs369956085 Chr.16: 50320226-50320226 on Build GRCh38
rs757240104 Chr.16: 50320256-50320256 on Build GRCh38
rs757663481 Chr.16: 50320258-50320258 on Build GRCh38
rs757591474 Chr.16: 50320327-50320327 on Build GRCh38
rs759525571 Chr.16: 50320304-50320304 on Build GRCh38
rs540625147 Chr.16: 50320392-50320392 on Build GRCh38
rs767793622 Chr.16: 50320262-50320262 on Build GRCh38
rs776525764 Chr.16: 50320302-50320302 on Build GRCh38
rs757898250 Chr.16: 50320371-50320371 on Build GRCh38
rs746419735 Chr.16: 50320274-50320274 on Build GRCh38
rs765295228 Chr.16: 50320307-50320307 on Build GRCh38
rs764381456 Chr.16: 50320253-50320253 on Build GRCh38
rs746626651 Chr.16: 50320383-50320383 on Build GRCh38
rs781513186 Chr.16: 50320272-50320272 on Build GRCh38
rs142511707 Chr.16: 50320318-50320318 on Build GRCh38
rs199797862 Chr.16: 50320395-50320395 on Build GRCh38
rs758685444 Chr.16: 50320235-50320235 on Build GRCh38
rs756618565 Chr.16: 50320361-50320361 on Build GRCh38
rs746738131 Chr.16: 50320299-50320299 on Build GRCh38
rs769781242 Chr.16: 50320406-50320406 on Build GRCh38
rs750977076 Chr.16: 50320333-50320333 on Build GRCh38
rs148031885 Chr.16: 50320294-50320294 on Build GRCh38
rs778627527 Chr.16: 50320285-50320285 on Build GRCh38
rs747802850 Chr.16: 50320293-50320293 on Build GRCh38
rs966140301 Chr.16: 50320336-50320336 on Build GRCh38
rs751814174 Chr.16: 50320254-50320254 on Build GRCh38
rs115491038 Chr.16: 50320312-50320312 on Build GRCh38
rs751773053 Chr.16: 50320324-50320324 on Build GRCh38
rs115672971 Chr.16: 50320389-50320389 on Build GRCh38
rs373486476 Chr.16: 50320279-50320279 on Build GRCh38
rs763089653 Chr.16: 50320399-50320400 on Build GRCh38
rs764271608 Chr.16: 50320321-50320321 on Build GRCh38
rs560321519 Chr.16: 50320408-50320408 on Build GRCh38
rs766612149 Chr.16: 50320398-50320398 on Build GRCh38
rs146033460 Chr.16: 50320329-50320329 on Build GRCh38
rs777324280 Chr.16: 50320380-50320380 on Build GRCh38
rs139953293 Chr.16: 50320369-50320369 on Build GRCh38
rs1056991255 Chr.16: 50320290-50320290 on Build GRCh38
rs780698234 Chr.16: 50320362-50320362 on Build GRCh38
rs1007854979 Chr.16: 50320311-50320311 on Build GRCh38
rs745658968 Chr.16: 50320398-50320398 on Build GRCh38
rs975754490 Chr.16: 50320354-50320354 on Build GRCh38

View Full Product Details