Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3849934-3850199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50B
Entrez Gene ID
26240
Gene Name
family with sequence similarity 50 member B
Gene Aliases
D6S2654E, X5L
UniGene
Hs.140944
Gene Chromosome Location
Chr.6: 3831901-3851320 on Build GRCh38
Transcripts
NM_012135.2 XM_017010729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659810 c.382G>A p.D128N Chr.6: 3850193-3850193 on Build GRCh38
3873632 c.256C>T p.R86W Chr.6: 3850067-3850067 on Build GRCh38
1444175 c.314G>A p.R105Q Chr.6: 3850125-3850125 on Build GRCh38
3873631 c.191C>T p.T64M Chr.6: 3850002-3850002 on Build GRCh38
6659806 c.250C>T p.R84C Chr.6: 3850061-3850061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774464980 Chr.6: 3850139-3850139 on Build GRCh38
rs775195740 Chr.6: 3850149-3850149 on Build GRCh38
rs117488732 Chr.6: 3850104-3850104 on Build GRCh38
rs144974260 Chr.6: 3849960-3849960 on Build GRCh38
rs755338605 Chr.6: 3850091-3850091 on Build GRCh38
rs777874858 Chr.6: 3850162-3850164 on Build GRCh38
rs763048166 Chr.6: 3850002-3850002 on Build GRCh38
rs759429994 Chr.6: 3850032-3850032 on Build GRCh38
rs748224989 Chr.6: 3849996-3849996 on Build GRCh38
rs747333793 Chr.6: 3850083-3850083 on Build GRCh38
rs766426175 Chr.6: 3850018-3850018 on Build GRCh38
rs752106061 Chr.6: 3850175-3850175 on Build GRCh38
rs773185779 Chr.6: 3849998-3849998 on Build GRCh38
rs376467863 Chr.6: 3850159-3850159 on Build GRCh38
rs767093077 Chr.6: 3849965-3849965 on Build GRCh38
rs774232202 Chr.6: 3850026-3850026 on Build GRCh38
rs139376471 Chr.6: 3849979-3849979 on Build GRCh38
rs758842799 Chr.6: 3850072-3850072 on Build GRCh38
rs771811466 Chr.6: 3850148-3850148 on Build GRCh38
rs760313884 Chr.6: 3850155-3850155 on Build GRCh38
rs763818446 Chr.6: 3850156-3850156 on Build GRCh38
rs149687084 Chr.6: 3849982-3849982 on Build GRCh38
rs147508307 Chr.6: 3849974-3849974 on Build GRCh38
rs754614628 Chr.6: 3849964-3849964 on Build GRCh38
rs763572373 Chr.6: 3850068-3850068 on Build GRCh38
rs758313883 Chr.6: 3850119-3850120 on Build GRCh38
rs777931418 Chr.6: 3850115-3850115 on Build GRCh38
rs769909140 Chr.6: 3850110-3850110 on Build GRCh38
rs552666804 Chr.6: 3850098-3850098 on Build GRCh38
rs766840357 Chr.6: 3850165-3850165 on Build GRCh38
rs770945816 Chr.6: 3850125-3850125 on Build GRCh38
rs913116353 Chr.6: 3850150-3850150 on Build GRCh38
rs749394834 Chr.6: 3850118-3850118 on Build GRCh38
rs753295618 Chr.6: 3850157-3850157 on Build GRCh38
rs145563282 Chr.6: 3849985-3849985 on Build GRCh38
rs866078475 Chr.6: 3849956-3849956 on Build GRCh38
rs148854528 Chr.6: 3850144-3850144 on Build GRCh38
rs755352309 Chr.6: 3850177-3850177 on Build GRCh38
rs755628628 Chr.6: 3850064-3850064 on Build GRCh38
rs767162341 Chr.6: 3850047-3850047 on Build GRCh38
rs746424924 Chr.6: 3849976-3849976 on Build GRCh38
rs780560493 Chr.6: 3850080-3850080 on Build GRCh38
rs752636834 Chr.6: 3850113-3850114 on Build GRCh38
rs370681792 Chr.6: 3850062-3850062 on Build GRCh38
rs1045601678 Chr.6: 3849961-3849961 on Build GRCh38
rs560129516 Chr.6: 3849973-3849973 on Build GRCh38
rs752206095 Chr.6: 3849966-3849966 on Build GRCh38
rs776749083 Chr.6: 3849991-3849991 on Build GRCh38
rs1028980548 Chr.6: 3850000-3850000 on Build GRCh38

View Full Product Details