Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36466043-36466316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF3R
Entrez Gene ID
1441
Gene Name
colony stimulating factor 3 receptor
Gene Aliases
CD114, GCSFR, SCN7
UniGene
Hs.524517
Gene Chromosome Location
Chr.1: 36466043-36483314 on Build GRCh38
Transcripts
NM_000760.3 NM_156039.3 NM_172313.2 XM_005270493.1 XM_011540748.2 XM_011540749.1 XM_011540750.1 XM_017000370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342325 c.2289C>A p.I763I Chr.1: 36466159-36466159 on Build GRCh38
464538 c.2249C>T p.A750V Chr.1: 36466199-36466199 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768258728 Chr.1: 36466210-36466210 on Build GRCh38
rs750397318 Chr.1: 36466145-36466145 on Build GRCh38
rs766114888 Chr.1: 36466161-36466161 on Build GRCh38
rs374816810 Chr.1: 36466228-36466228 on Build GRCh38
rs992016941 Chr.1: 36466222-36466222 on Build GRCh38
rs779336097 Chr.1: 36466194-36466194 on Build GRCh38
rs931211255 Chr.1: 36466185-36466185 on Build GRCh38
rs772439953 Chr.1: 36466088-36466088 on Build GRCh38
rs142939584 Chr.1: 36466200-36466200 on Build GRCh38
rs367706643 Chr.1: 36466177-36466177 on Build GRCh38
rs563278395 Chr.1: 36466283-36466283 on Build GRCh38
rs368221209 Chr.1: 36466236-36466236 on Build GRCh38
rs199810615 Chr.1: 36466135-36466135 on Build GRCh38
rs764841061 Chr.1: 36466231-36466231 on Build GRCh38
rs772443391 Chr.1: 36466196-36466196 on Build GRCh38
rs752919171 Chr.1: 36466174-36466174 on Build GRCh38
rs764036081 Chr.1: 36466086-36466086 on Build GRCh38
rs760266886 Chr.1: 36466109-36466109 on Build GRCh38
rs774355214 Chr.1: 36466107-36466107 on Build GRCh38
rs372314097 Chr.1: 36466251-36466251 on Build GRCh38
rs776042988 Chr.1: 36466124-36466124 on Build GRCh38
rs771983075 Chr.1: 36466256-36466256 on Build GRCh38
rs776266691 Chr.1: 36466212-36466212 on Build GRCh38
rs144368887 Chr.1: 36466281-36466281 on Build GRCh38
rs895899819 Chr.1: 36466254-36466254 on Build GRCh38
rs775142640 Chr.1: 36466093-36466093 on Build GRCh38
rs575585504 Chr.1: 36466288-36466288 on Build GRCh38
rs370378939 Chr.1: 36466203-36466203 on Build GRCh38
rs367919345 Chr.1: 36466160-36466160 on Build GRCh38
rs1020453765 Chr.1: 36466263-36466263 on Build GRCh38
rs190798269 Chr.1: 36466127-36466127 on Build GRCh38
rs777880216 Chr.1: 36466184-36466184 on Build GRCh38
rs758195954 Chr.1: 36466152-36466152 on Build GRCh38
rs1048283841 Chr.1: 36466191-36466191 on Build GRCh38
rs775765229 Chr.1: 36466198-36466198 on Build GRCh38
rs534697692 Chr.1: 36466113-36466113 on Build GRCh38

View Full Product Details