Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40849749-40850010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A6
Entrez Gene ID
1548
Gene Name
cytochrome P450 family 2 subfamily A member 6
Gene Aliases
CPA6, CYP2A, CYP2A3, CYPIIA6, P450C2A, P450PB
UniGene
Hs.250615
Gene Chromosome Location
Chr.19: 40843538-40850447 on Build GRCh38
Transcripts
NM_000762.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
215771 c.302G>A p.R101Q Chr.19: 40849859-40849859 on Build GRCh38
996850 c.191G>A p.R64H Chr.19: 40849970-40849970 on Build GRCh38
996849 c.235G>A p.V79M Chr.19: 40849926-40849926 on Build GRCh38
6367611 c.227G>A p.R76Q Chr.19: 40849934-40849934 on Build GRCh38
5364974 c.193T>C p.Y65H Chr.19: 40849968-40849968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774455621 Chr.19: 40849824-40849824 on Build GRCh38
rs72549434 Chr.19: 40849849-40849849 on Build GRCh38
rs372082998 Chr.19: 40849898-40849898 on Build GRCh38
rs377137055 Chr.19: 40849776-40849776 on Build GRCh38
rs768460099 Chr.19: 40849793-40849793 on Build GRCh38
rs766889674 Chr.19: 40849896-40849896 on Build GRCh38
rs147022782 Chr.19: 40849927-40849927 on Build GRCh38
rs770128266 Chr.19: 40849984-40849984 on Build GRCh38
rs769660507 Chr.19: 40849801-40849801 on Build GRCh38
rs772141829 Chr.19: 40849880-40849880 on Build GRCh38
rs142692109 Chr.19: 40849920-40849920 on Build GRCh38
rs763613869 Chr.19: 40849932-40849932 on Build GRCh38
rs747840206 Chr.19: 40849862-40849862 on Build GRCh38
rs752996917 Chr.19: 40849834-40849834 on Build GRCh38
rs143067113 Chr.19: 40849902-40849902 on Build GRCh38
rs753492338 Chr.19: 40849958-40849958 on Build GRCh38
rs764536563 Chr.19: 40849840-40849840 on Build GRCh38
rs759427328 Chr.19: 40849876-40849876 on Build GRCh38
rs762249658 Chr.19: 40849896-40849896 on Build GRCh38
rs760744271 Chr.19: 40849839-40849839 on Build GRCh38
rs754157454 Chr.19: 40849989-40849989 on Build GRCh38
rs149065015 Chr.19: 40849884-40849884 on Build GRCh38
rs754143940 Chr.19: 40849847-40849847 on Build GRCh38
rs747024492 Chr.19: 40849926-40849926 on Build GRCh38
rs752135850 Chr.19: 40849904-40849904 on Build GRCh38
rs537450108 Chr.19: 40849805-40849805 on Build GRCh38
rs776709300 Chr.19: 40849979-40849979 on Build GRCh38
rs370568696 Chr.19: 40849782-40849782 on Build GRCh38
rs777681590 Chr.19: 40849864-40849864 on Build GRCh38
rs368359507 Chr.19: 40849897-40849897 on Build GRCh38
rs775702923 Chr.19: 40849924-40849924 on Build GRCh38
rs759590848 Chr.19: 40849828-40849828 on Build GRCh38
rs747704342 Chr.19: 40849799-40849799 on Build GRCh38
rs770841973 Chr.19: 40849822-40849822 on Build GRCh38
rs546628995 Chr.19: 40849796-40849796 on Build GRCh38
rs762090075 Chr.19: 40849930-40849930 on Build GRCh38
rs72549436 Chr.19: 40849771-40849771 on Build GRCh38
rs199545200 Chr.19: 40849860-40849860 on Build GRCh38
rs757073282 Chr.19: 40849914-40849914 on Build GRCh38
rs776744913 Chr.19: 40849929-40849929 on Build GRCh38
rs777268861 Chr.19: 40849911-40849911 on Build GRCh38
rs777098140 Chr.19: 40849887-40849887 on Build GRCh38
rs28609092 Chr.19: 40849870-40849870 on Build GRCh38
rs201833760 Chr.19: 40849933-40849933 on Build GRCh38
rs772814182 Chr.19: 40849816-40849816 on Build GRCh38
rs948874490 Chr.19: 40849935-40849935 on Build GRCh38
rs771420298 Chr.19: 40849813-40849813 on Build GRCh38
rs767705968 Chr.19: 40849775-40849776 on Build GRCh38
rs769272500 Chr.19: 40849863-40849863 on Build GRCh38
rs199702575 Chr.19: 40849915-40849915 on Build GRCh38
rs143690364 Chr.19: 40849959-40849959 on Build GRCh38
rs746509603 Chr.19: 40849788-40849788 on Build GRCh38
rs752300065 Chr.19: 40849934-40849934 on Build GRCh38
rs750510845 Chr.19: 40849778-40849779 on Build GRCh38
rs375051929 Chr.19: 40849859-40849859 on Build GRCh38
rs72549433 Chr.19: 40849960-40849960 on Build GRCh38
rs77233796 Chr.19: 40849923-40849923 on Build GRCh38
rs373775804 Chr.19: 40849773-40849773 on Build GRCh38
rs182169150 Chr.19: 40849819-40849819 on Build GRCh38
rs763618910 Chr.19: 40849957-40849957 on Build GRCh38
rs764214264 Chr.19: 40849885-40849885 on Build GRCh38
rs201250423 Chr.19: 40849931-40849931 on Build GRCh38
rs758397496 Chr.19: 40849966-40849966 on Build GRCh38
rs755862580 Chr.19: 40849910-40849910 on Build GRCh38
rs777961854 Chr.19: 40849775-40849775 on Build GRCh38
rs2302990 Chr.19: 40849944-40849944 on Build GRCh38
rs1015296582 Chr.19: 40849804-40849804 on Build GRCh38
rs779527856 Chr.19: 40849855-40849855 on Build GRCh38
rs201550898 Chr.19: 40849903-40849903 on Build GRCh38
rs199515342 Chr.19: 40849971-40849971 on Build GRCh38
rs745654874 Chr.19: 40849962-40849962 on Build GRCh38
rs577267650 Chr.19: 40849881-40849881 on Build GRCh38
rs148202548 Chr.19: 40849912-40849912 on Build GRCh38
rs749005083 Chr.19: 40849866-40849866 on Build GRCh38
rs376970015 Chr.19: 40849991-40849991 on Build GRCh38
rs28399439 Chr.19: 40849809-40849809 on Build GRCh38
rs200554095 Chr.19: 40849980-40849980 on Build GRCh38
rs72549435 Chr.19: 40849833-40849833 on Build GRCh38
rs374515279 Chr.19: 40849970-40849970 on Build GRCh38
rs145308399 Chr.19: 40849872-40849872 on Build GRCh38

View Full Product Details