Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43576974-43577219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO10
Entrez Gene ID
55129
Gene Name
anoctamin 10
Gene Aliases
SCAR10, TMEM16K
UniGene
Hs.656657
Gene Chromosome Location
Chr.3: 43366324-43691594 on Build GRCh38
Transcripts
NM_001204831.1 NM_001204832.1 NM_001204833.1 NM_001204834.1 NM_018075.3 XM_011533885.2 XM_011533887.2 XM_011533889.2 XM_011533890.2 XM_017006716.1 XM_017006717.1 XM_017006718.1 XM_017006719.1 XM_017006720.1 XM_017006721.1 XM_017006722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7151145 c.731T>C p.V244A Chr.3: 43577123-43577123 on Build GRCh38
6010945 c.693T>C p.P231P Chr.3: 43577161-43577161 on Build GRCh38
6534982 c.719A>C p.Y240S Chr.3: 43577135-43577135 on Build GRCh38
5355697 c.759G>T p.W253C Chr.3: 43577095-43577095 on Build GRCh38
7184338 c.865C>G p.H289D Chr.3: 43576989-43576989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753407149 Chr.3: 43577101-43577101 on Build GRCh38
rs372215383 Chr.3: 43577052-43577052 on Build GRCh38
rs369488032 Chr.3: 43577067-43577067 on Build GRCh38
rs776868518 Chr.3: 43577045-43577045 on Build GRCh38
rs188443010 Chr.3: 43577178-43577178 on Build GRCh38
rs750170555 Chr.3: 43577109-43577110 on Build GRCh38
rs763560921 Chr.3: 43577186-43577186 on Build GRCh38
rs774238439 Chr.3: 43577188-43577188 on Build GRCh38
rs759828781 Chr.3: 43577168-43577168 on Build GRCh38
rs752208261 Chr.3: 43577089-43577089 on Build GRCh38
rs769678648 Chr.3: 43577000-43577000 on Build GRCh38
rs565581904 Chr.3: 43577113-43577113 on Build GRCh38
rs201052710 Chr.3: 43577124-43577124 on Build GRCh38
rs766344882 Chr.3: 43577172-43577172 on Build GRCh38
rs531102308 Chr.3: 43577116-43577116 on Build GRCh38
rs200840765 Chr.3: 43577076-43577076 on Build GRCh38
rs369417903 Chr.3: 43577122-43577122 on Build GRCh38
rs201848805 Chr.3: 43576999-43576999 on Build GRCh38
rs772633592 Chr.3: 43577171-43577171 on Build GRCh38
rs201491185 Chr.3: 43577111-43577111 on Build GRCh38
rs373217121 Chr.3: 43577110-43577110 on Build GRCh38
rs756881754 Chr.3: 43576999-43576999 on Build GRCh38
rs143322256 Chr.3: 43577055-43577055 on Build GRCh38
rs184756780 Chr.3: 43577012-43577012 on Build GRCh38
rs756986703 Chr.3: 43577019-43577019 on Build GRCh38
rs760224425 Chr.3: 43577185-43577185 on Build GRCh38
rs763793134 Chr.3: 43577098-43577098 on Build GRCh38
rs749710737 Chr.3: 43577147-43577147 on Build GRCh38
rs374795191 Chr.3: 43577017-43577017 on Build GRCh38
rs773557659 Chr.3: 43577062-43577062 on Build GRCh38
rs767044966 Chr.3: 43577001-43577001 on Build GRCh38
rs375157457 Chr.3: 43576997-43576997 on Build GRCh38
rs778539546 Chr.3: 43577103-43577103 on Build GRCh38
rs771135471 Chr.3: 43577148-43577148 on Build GRCh38
rs772092273 Chr.3: 43577039-43577039 on Build GRCh38
rs375470443 Chr.3: 43577064-43577064 on Build GRCh38
rs41289586 Chr.3: 43577066-43577066 on Build GRCh38
rs367711525 Chr.3: 43577021-43577021 on Build GRCh38
rs780140084 Chr.3: 43577114-43577114 on Build GRCh38
rs767953054 Chr.3: 43577082-43577082 on Build GRCh38
rs192531429 Chr.3: 43577004-43577004 on Build GRCh38
rs757113016 Chr.3: 43577104-43577104 on Build GRCh38
rs755615560 Chr.3: 43577090-43577090 on Build GRCh38
rs868729445 Chr.3: 43577026-43577026 on Build GRCh38

View Full Product Details