Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73959431-73959704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB13
Entrez Gene ID
374407
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Gene Aliases
RSPH16A, TSARG5, TSARG6
UniGene
Hs.567888
Gene Chromosome Location
Chr.11: 73950319-73970287 on Build GRCh38
Transcripts
NM_153614.3 XM_005273984.3 XM_011545004.2 XM_011545005.2 XM_011545007.2 XM_011545009.2 XM_011545013.2 XM_011545014.2 XM_017017675.1 XM_017017676.1 XM_017017677.1 XM_017017678.1 XM_017017679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5839581 c.293A>G p.K98R Chr.11: 73959614-73959614 on Build GRCh38
4517362 c.326_327CC>TT p.P109L Chr.11: 73959647-73959648 on Build GRCh38
5812124 c.287C>T p.P96L Chr.11: 73959608-73959608 on Build GRCh38
274844 c.309C>A p.F103L Chr.11: 73959630-73959630 on Build GRCh38
1204176 c.197A>C p.K66T Chr.11: 73959518-73959518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036679359 Chr.11: 73959459-73959459 on Build GRCh38
rs781143290 Chr.11: 73959481-73959481 on Build GRCh38
rs759212996 Chr.11: 73959471-73959473 on Build GRCh38
rs653263 Chr.11: 73959600-73959600 on Build GRCh38
rs765697242 Chr.11: 73959564-73959564 on Build GRCh38
rs756043336 Chr.11: 73959617-73959617 on Build GRCh38
rs267603184 Chr.11: 73959546-73959546 on Build GRCh38
rs750950916 Chr.11: 73959550-73959550 on Build GRCh38
rs755920594 Chr.11: 73959490-73959490 on Build GRCh38
rs757230313 Chr.11: 73959477-73959477 on Build GRCh38
rs771877900 Chr.11: 73959548-73959558 on Build GRCh38
rs199638499 Chr.11: 73959676-73959676 on Build GRCh38
rs1022933231 Chr.11: 73959584-73959584 on Build GRCh38
rs777879695 Chr.11: 73959658-73959658 on Build GRCh38
rs762711919 Chr.11: 73959591-73959591 on Build GRCh38
rs368613359 Chr.11: 73959513-73959513 on Build GRCh38
rs771084987 Chr.11: 73959525-73959525 on Build GRCh38
rs747116650 Chr.11: 73959522-73959522 on Build GRCh38
rs145909750 Chr.11: 73959593-73959593 on Build GRCh38
rs750960066 Chr.11: 73959648-73959648 on Build GRCh38
rs747168118 Chr.11: 73959666-73959666 on Build GRCh38
rs999344777 Chr.11: 73959501-73959501 on Build GRCh38
rs754762238 Chr.11: 73959655-73959655 on Build GRCh38
rs1053124103 Chr.11: 73959504-73959504 on Build GRCh38
rs775701724 Chr.11: 73959577-73959577 on Build GRCh38
rs200540532 Chr.11: 73959592-73959592 on Build GRCh38
rs995414815 Chr.11: 73959500-73959500 on Build GRCh38
rs898202211 Chr.11: 73959484-73959484 on Build GRCh38
rs577291859 Chr.11: 73959665-73959665 on Build GRCh38
rs201272687 Chr.11: 73959514-73959514 on Build GRCh38
rs375146507 Chr.11: 73959505-73959505 on Build GRCh38
rs376769428 Chr.11: 73959566-73959566 on Build GRCh38
rs1019878020 Chr.11: 73959675-73959675 on Build GRCh38
rs777289652 Chr.11: 73959542-73959542 on Build GRCh38
rs186414805 Chr.11: 73959674-73959674 on Build GRCh38
rs751008022 Chr.11: 73959457-73959457 on Build GRCh38
rs750366252 Chr.11: 73959616-73959616 on Build GRCh38
rs140841219 Chr.11: 73959506-73959506 on Build GRCh38
rs372254426 Chr.11: 73959517-73959517 on Build GRCh38
rs763713198 Chr.11: 73959456-73959456 on Build GRCh38
rs766959001 Chr.11: 73959601-73959601 on Build GRCh38
rs375920351 Chr.11: 73959668-73959668 on Build GRCh38
rs368904856 Chr.11: 73959624-73959624 on Build GRCh38
rs776842427 Chr.11: 73959672-73959672 on Build GRCh38

View Full Product Details