Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4901911-4902138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf107
Entrez Gene ID
100130311
Gene Name
chromosome 17 open reading frame 107
Gene Aliases
-
UniGene
Hs.579243
Gene Chromosome Location
Chr.17: 4899653-4902932 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628805 c.434G>A p.R145H Chr.17: 4901998-4901998 on Build GRCh38
5364417 c.399C>T p.G133G Chr.17: 4902033-4902033 on Build GRCh38
6628804 c.439G>A p.V147I Chr.17: 4901993-4901993 on Build GRCh38
6030765 c.376G>A p.A126T Chr.17: 4902056-4902056 on Build GRCh38
4970160 c.367G>C p.A123P Chr.17: 4902065-4902065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751665448 Chr.17: 4902033-4902033 on Build GRCh38
rs760043533 Chr.17: 4902085-4902085 on Build GRCh38
rs752818051 Chr.17: 4902046-4902046 on Build GRCh38
rs751830305 Chr.17: 4901964-4901964 on Build GRCh38
rs750257362 Chr.17: 4902100-4902100 on Build GRCh38
rs777324821 Chr.17: 4902036-4902037 on Build GRCh38
rs771950733 Chr.17: 4901993-4901993 on Build GRCh38
rs779058823 Chr.17: 4901991-4901991 on Build GRCh38
rs747638974 Chr.17: 4902075-4902075 on Build GRCh38
rs770978243 Chr.17: 4901984-4901984 on Build GRCh38
rs375169047 Chr.17: 4902091-4902091 on Build GRCh38
rs758084108 Chr.17: 4902057-4902057 on Build GRCh38
rs1056080125 Chr.17: 4902059-4902059 on Build GRCh38
rs754300536 Chr.17: 4901952-4901952 on Build GRCh38
rs121909514 Chr.17: 4901932-4901932 on Build GRCh38
rs758366677 Chr.17: 4902104-4902104 on Build GRCh38
rs780779198 Chr.17: 4902107-4902107 on Build GRCh38
rs121909512 Chr.17: 4902010-4902010 on Build GRCh38
rs769318953 Chr.17: 4902076-4902076 on Build GRCh38
rs886038226 Chr.17: 4902012-4902012 on Build GRCh38
rs766798452 Chr.17: 4902030-4902030 on Build GRCh38
rs773553639 Chr.17: 4902077-4902077 on Build GRCh38
rs756213541 Chr.17: 4902098-4902098 on Build GRCh38
rs985566505 Chr.17: 4902117-4902117 on Build GRCh38
rs756301699 Chr.17: 4902051-4902051 on Build GRCh38
rs140042356 Chr.17: 4902072-4902072 on Build GRCh38
rs765794362 Chr.17: 4901953-4901953 on Build GRCh38
rs771356927 Chr.17: 4902073-4902073 on Build GRCh38
rs369060595 Chr.17: 4902099-4902099 on Build GRCh38
rs368846437 Chr.17: 4901963-4901963 on Build GRCh38
rs535287601 Chr.17: 4901975-4901975 on Build GRCh38
rs183983159 Chr.17: 4902032-4902032 on Build GRCh38
rs773822199 Chr.17: 4902026-4902026 on Build GRCh38
rs139625105 Chr.17: 4901967-4901967 on Build GRCh38
rs778196743 Chr.17: 4901982-4901982 on Build GRCh38
rs72835059 Chr.17: 4902094-4902094 on Build GRCh38
rs142901746 Chr.17: 4901977-4901977 on Build GRCh38
rs146089157 Chr.17: 4902022-4902022 on Build GRCh38
rs781427923 Chr.17: 4901970-4901970 on Build GRCh38
rs970593355 Chr.17: 4901988-4901988 on Build GRCh38
rs776429372 Chr.17: 4901994-4901994 on Build GRCh38
rs763520880 Chr.17: 4902078-4902078 on Build GRCh38
rs749659243 Chr.17: 4902042-4902042 on Build GRCh38
rs199708479 Chr.17: 4902083-4902083 on Build GRCh38
rs201555272 Chr.17: 4902106-4902106 on Build GRCh38
rs763181728 Chr.17: 4902028-4902028 on Build GRCh38
rs750784391 Chr.17: 4901954-4901954 on Build GRCh38
rs894382905 Chr.17: 4902060-4902060 on Build GRCh38
rs200515887 Chr.17: 4902056-4902056 on Build GRCh38
rs571215814 Chr.17: 4902055-4902055 on Build GRCh38
rs483353045 Chr.17: 4902011-4902011 on Build GRCh38
rs746582621 Chr.17: 4902070-4902070 on Build GRCh38
rs766595838 Chr.17: 4902082-4902082 on Build GRCh38
rs121909516 Chr.17: 4901944-4901944 on Build GRCh38
rs958078989 Chr.17: 4902050-4902050 on Build GRCh38
rs760402668 Chr.17: 4902007-4902007 on Build GRCh38
rs747832866 Chr.17: 4902119-4902119 on Build GRCh38
rs767908282 Chr.17: 4902045-4902045 on Build GRCh38
rs775247070 Chr.17: 4901933-4901933 on Build GRCh38
rs150868720 Chr.17: 4901958-4901958 on Build GRCh38
rs779901038 Chr.17: 4902069-4902069 on Build GRCh38
rs776129600 Chr.17: 4902017-4902017 on Build GRCh38

View Full Product Details