Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119697842-119698097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIT
Entrez Gene ID
11113
Gene Name
citron rho-interacting serine/threonine kinase
Gene Aliases
CRIK, STK21
UniGene
Hs.119594
Gene Chromosome Location
Chr.12: 119685790-119877318 on Build GRCh38
Transcripts
NM_001206999.1 NM_007174.2 XM_006719206.2 XM_011537783.1 XM_011537784.1 XM_011537785.1 XM_011537787.1 XM_011537788.1 XM_011537789.1 XM_011537790.2 XM_011537791.2 XM_011537792.2 XM_017018735.1 XM_017018736.1 XM_017018737.1 XM_017018738.1 XM_017018739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629354 c.5571A>G p.S1857S Chr.12: 119697981-119697981 on Build GRCh38
4973632 c.5563C>T p.R1855C Chr.12: 119697989-119697989 on Build GRCh38
3980815 c.5539C>G p.L1847V Chr.12: 119698013-119698013 on Build GRCh38
3456911 c.5567C>T p.S1856F Chr.12: 119697985-119697985 on Build GRCh38
7003763 c.5535C>G p.N1845K Chr.12: 119698017-119698017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770428023 Chr.12: 119697989-119697989 on Build GRCh38
rs145997090 Chr.12: 119697954-119697954 on Build GRCh38
rs528707936 Chr.12: 119697960-119697960 on Build GRCh38
rs772698952 Chr.12: 119697871-119697871 on Build GRCh38
rs542463927 Chr.12: 119697961-119697961 on Build GRCh38
rs780026255 Chr.12: 119697955-119697955 on Build GRCh38
rs577845147 Chr.12: 119697940-119697940 on Build GRCh38
rs761177635 Chr.12: 119698027-119698027 on Build GRCh38
rs769416804 Chr.12: 119697966-119697966 on Build GRCh38
rs1036824380 Chr.12: 119698025-119698025 on Build GRCh38
rs543886772 Chr.12: 119697914-119697914 on Build GRCh38
rs278126 Chr.12: 119697912-119697912 on Build GRCh38
rs767880962 Chr.12: 119697876-119697876 on Build GRCh38
rs974242303 Chr.12: 119697920-119697920 on Build GRCh38
rs769506702 Chr.12: 119697870-119697870 on Build GRCh38
rs1054955956 Chr.12: 119698078-119698078 on Build GRCh38
rs779334610 Chr.12: 119697952-119697952 on Build GRCh38
rs762315082 Chr.12: 119698051-119698051 on Build GRCh38
rs372312938 Chr.12: 119697925-119697925 on Build GRCh38
rs375339875 Chr.12: 119697987-119697987 on Build GRCh38
rs199807885 Chr.12: 119697958-119697958 on Build GRCh38
rs754240898 Chr.12: 119698050-119698050 on Build GRCh38
rs762502026 Chr.12: 119697873-119697873 on Build GRCh38
rs763157071 Chr.12: 119697988-119697988 on Build GRCh38
rs115111326 Chr.12: 119698077-119698077 on Build GRCh38
rs765523938 Chr.12: 119698063-119698063 on Build GRCh38
rs757547985 Chr.12: 119697941-119697941 on Build GRCh38
rs753209630 Chr.12: 119697880-119697880 on Build GRCh38
rs750630445 Chr.12: 119698071-119698071 on Build GRCh38
rs761108605 Chr.12: 119697886-119697886 on Build GRCh38
rs768487793 Chr.12: 119697959-119697959 on Build GRCh38
rs764574266 Chr.12: 119698029-119698029 on Build GRCh38
rs773747883 Chr.12: 119698011-119698011 on Build GRCh38
rs962468884 Chr.12: 119697918-119697918 on Build GRCh38

View Full Product Details