Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111882165-111882438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf1
Entrez Gene ID
64776
Gene Name
chromosome 11 open reading frame 1
Gene Aliases
-
UniGene
Hs.17546
Gene Chromosome Location
Chr.11: 111879190-111884083 on Build GRCh38
Transcripts
NM_022761.2 XM_011542957.2 XM_011542958.2 XM_017018129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4912759 c.53A>T p.N18I Chr.11: 111882375-111882375 on Build GRCh38
5421004 c.67C>T p.L23F Chr.11: 111882389-111882389 on Build GRCh38
4574043 c.57C>T p.L19L Chr.11: 111882379-111882379 on Build GRCh38
5925552 c.46-5C>T p.? Chr.11: 111882363-111882363 on Build GRCh38
4666585 c.58G>A p.A20T Chr.11: 111882380-111882380 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372958672 Chr.11: 111882373-111882373 on Build GRCh38
rs935455158 Chr.11: 111882226-111882226 on Build GRCh38
rs782008618 Chr.11: 111882285-111882285 on Build GRCh38
rs782211356 Chr.11: 111882393-111882394 on Build GRCh38
rs782330820 Chr.11: 111882284-111882284 on Build GRCh38
rs782148488 Chr.11: 111882415-111882416 on Build GRCh38
rs782789146 Chr.11: 111882382-111882382 on Build GRCh38
rs782633128 Chr.11: 111882236-111882236 on Build GRCh38
rs568239268 Chr.11: 111882300-111882300 on Build GRCh38
rs376348016 Chr.11: 111882398-111882398 on Build GRCh38
rs782180413 Chr.11: 111882263-111882263 on Build GRCh38
rs782296273 Chr.11: 111882349-111882349 on Build GRCh38
rs568806168 Chr.11: 111882365-111882365 on Build GRCh38
rs781965856 Chr.11: 111882384-111882385 on Build GRCh38
rs200607083 Chr.11: 111882347-111882347 on Build GRCh38
rs369117585 Chr.11: 111882370-111882370 on Build GRCh38
rs782300733 Chr.11: 111882376-111882376 on Build GRCh38
rs781923077 Chr.11: 111882379-111882379 on Build GRCh38
rs924042475 Chr.11: 111882204-111882204 on Build GRCh38
rs782042217 Chr.11: 111882404-111882405 on Build GRCh38
rs1038929848 Chr.11: 111882348-111882348 on Build GRCh38
rs552282748 Chr.11: 111882287-111882287 on Build GRCh38
rs782110008 Chr.11: 111882392-111882392 on Build GRCh38
rs915281453 Chr.11: 111882293-111882293 on Build GRCh38
rs535230725 Chr.11: 111882304-111882304 on Build GRCh38
rs139660891 Chr.11: 111882380-111882380 on Build GRCh38
rs781819447 Chr.11: 111882411-111882411 on Build GRCh38
rs782495122 Chr.11: 111882412-111882412 on Build GRCh38
rs782375655 Chr.11: 111882350-111882350 on Build GRCh38
rs376776600 Chr.11: 111882384-111882384 on Build GRCh38
rs782785229 Chr.11: 111882394-111882394 on Build GRCh38
rs782331093 Chr.11: 111882337-111882337 on Build GRCh38

View Full Product Details