Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111248508-111248769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN6
Entrez Gene ID
827
Gene Name
calpain 6
Gene Aliases
CANPX, CAPNX, CalpM, DJ914P14.1
UniGene
Hs.496593
Gene Chromosome Location
Chr.X: 111245099-111270546 on Build GRCh38
Transcripts
NM_014289.3

SNP Detail

SNP ID Chromosome Location
rs961625582 Chr.X: 111248697-111248697 on Build GRCh38
rs781222052 Chr.X: 111248619-111248619 on Build GRCh38
rs17879776 Chr.X: 111248637-111248637 on Build GRCh38
rs762409717 Chr.X: 111248539-111248539 on Build GRCh38
rs768150366 Chr.X: 111248650-111248650 on Build GRCh38
rs1035507245 Chr.X: 111248642-111248642 on Build GRCh38
rs773773647 Chr.X: 111248706-111248706 on Build GRCh38
rs766496943 Chr.X: 111248723-111248723 on Build GRCh38
rs757446645 Chr.X: 111248554-111248554 on Build GRCh38
rs772570297 Chr.X: 111248688-111248688 on Build GRCh38
rs369291940 Chr.X: 111248625-111248625 on Build GRCh38
rs760680053 Chr.X: 111248559-111248559 on Build GRCh38
rs779685988 Chr.X: 111248626-111248626 on Build GRCh38
rs745378811 Chr.X: 111248621-111248621 on Build GRCh38
rs755136186 Chr.X: 111248589-111248589 on Build GRCh38
rs147998864 Chr.X: 111248604-111248604 on Build GRCh38
rs559083969 Chr.X: 111248603-111248603 on Build GRCh38
rs1023858527 Chr.X: 111248579-111248579 on Build GRCh38
rs200492199 Chr.X: 111248547-111248547 on Build GRCh38
rs751274083 Chr.X: 111248609-111248609 on Build GRCh38
rs774799400 Chr.X: 111248656-111248656 on Build GRCh38
rs754118670 Chr.X: 111248572-111248572 on Build GRCh38
rs776542662 Chr.X: 111248732-111248732 on Build GRCh38
rs1003743848 Chr.X: 111248628-111248628 on Build GRCh38
rs1011083423 Chr.X: 111248568-111248568 on Build GRCh38
rs372641764 Chr.X: 111248674-111248674 on Build GRCh38
rs951790817 Chr.X: 111248555-111248555 on Build GRCh38
rs766441218 Chr.X: 111248571-111248571 on Build GRCh38
rs761158567 Chr.X: 111248713-111248713 on Build GRCh38
rs200048224 Chr.X: 111248614-111248614 on Build GRCh38

View Full Product Details