Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93259507-93259758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS50
Entrez Gene ID
55610
Gene Name
VPS50, EARP/GARPII complex subunit
Gene Aliases
CCDC132, VPS54L
UniGene
Hs.202424
Gene Chromosome Location
Chr.7: 93232340-93361123 on Build GRCh38
Transcripts
NM_001257998.1 NM_017667.3 NM_024553.2 XM_011516395.2 XM_011516396.2

SNP Detail

SNP ID Chromosome Location
rs768541908 Chr.7: 93259539-93259539 on Build GRCh38
rs936197006 Chr.7: 93259688-93259688 on Build GRCh38
rs754378557 Chr.7: 93259638-93259638 on Build GRCh38
rs768453681 Chr.7: 93259666-93259666 on Build GRCh38
rs767959681 Chr.7: 93259593-93259593 on Build GRCh38
rs758088966 Chr.7: 93259661-93259661 on Build GRCh38
rs746758245 Chr.7: 93259663-93259663 on Build GRCh38
rs977658370 Chr.7: 93259656-93259656 on Build GRCh38
rs552294557 Chr.7: 93259703-93259703 on Build GRCh38
rs745488255 Chr.7: 93259658-93259658 on Build GRCh38
rs759849081 Chr.7: 93259580-93259580 on Build GRCh38
rs776546133 Chr.7: 93259671-93259671 on Build GRCh38
rs771935998 Chr.7: 93259574-93259574 on Build GRCh38
rs779714100 Chr.7: 93259662-93259662 on Build GRCh38
rs756566977 Chr.7: 93259608-93259608 on Build GRCh38
rs367907923 Chr.7: 93259538-93259538 on Build GRCh38
rs200949909 Chr.7: 93259570-93259570 on Build GRCh38
rs372089506 Chr.7: 93259622-93259622 on Build GRCh38
rs762121374 Chr.7: 93259641-93259646 on Build GRCh38
rs762874399 Chr.7: 93259564-93259564 on Build GRCh38
rs764634283 Chr.7: 93259614-93259614 on Build GRCh38
rs776572775 Chr.7: 93259542-93259542 on Build GRCh38
rs748173864 Chr.7: 93259678-93259678 on Build GRCh38
rs756721124 Chr.7: 93259649-93259649 on Build GRCh38
rs753067603 Chr.7: 93259607-93259607 on Build GRCh38
rs766547007 Chr.7: 93259568-93259568 on Build GRCh38
rs1018619202 Chr.7: 93259603-93259603 on Build GRCh38
rs926111864 Chr.7: 93259672-93259672 on Build GRCh38

View Full Product Details