Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17199348-17199621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR1
Entrez Gene ID
51816
Gene Name
cat eye syndrome chromosome region, candidate 1
Gene Aliases
ADA2, ADGF, IDGFL, PAN, SNEDS
UniGene
Hs.170310
Gene Chromosome Location
Chr.22: 17178790-17221854 on Build GRCh38
Transcripts
NM_001282225.1 NM_001282226.1 NM_001282227.1 NM_001282228.1 NM_001282229.1 NM_177405.2 XM_006724080.3 XM_011546133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445630 c.30+3A>T p.? Chr.22: 17199546-17199546 on Build GRCh38
6559238 c.4G>A p.D2N Chr.22: 17199575-17199575 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs975819397 Chr.22: 17199389-17199389 on Build GRCh38
rs367818922 Chr.22: 17199511-17199511 on Build GRCh38
rs778288216 Chr.22: 17199509-17199509 on Build GRCh38
rs760865260 Chr.22: 17199589-17199590 on Build GRCh38
rs751908953 Chr.22: 17199588-17199588 on Build GRCh38
rs868071458 Chr.22: 17199442-17199442 on Build GRCh38
rs564924449 Chr.22: 17199495-17199495 on Build GRCh38
rs781299779 Chr.22: 17199525-17199525 on Build GRCh38
rs749513715 Chr.22: 17199553-17199553 on Build GRCh38
rs71200246 Chr.22: 17199448-17199449 on Build GRCh38
rs766270323 Chr.22: 17199571-17199571 on Build GRCh38
rs764514432 Chr.22: 17199587-17199587 on Build GRCh38
rs572953053 Chr.22: 17199462-17199462 on Build GRCh38
rs540274197 Chr.22: 17199477-17199477 on Build GRCh38
rs557300400 Chr.22: 17199371-17199371 on Build GRCh38
rs577737169 Chr.22: 17199376-17199376 on Build GRCh38
rs539595665 Chr.22: 17199551-17199551 on Build GRCh38
rs769883211 Chr.22: 17199545-17199545 on Build GRCh38
rs771408477 Chr.22: 17199521-17199521 on Build GRCh38
rs144627322 Chr.22: 17199416-17199417 on Build GRCh38
rs773289382 Chr.22: 17199535-17199536 on Build GRCh38
rs544476565 Chr.22: 17199549-17199549 on Build GRCh38
rs745982014 Chr.22: 17199534-17199534 on Build GRCh38
rs368098668 Chr.22: 17199376-17199377 on Build GRCh38
rs776185534 Chr.22: 17199572-17199572 on Build GRCh38
rs922842729 Chr.22: 17199430-17199430 on Build GRCh38
rs772934083 Chr.22: 17199570-17199570 on Build GRCh38
rs771792628 Chr.22: 17199556-17199556 on Build GRCh38
rs757759335 Chr.22: 17199596-17199596 on Build GRCh38
rs189153315 Chr.22: 17199465-17199465 on Build GRCh38
rs934278685 Chr.22: 17199467-17199467 on Build GRCh38
rs758138377 Chr.22: 17199584-17199584 on Build GRCh38
rs562613700 Chr.22: 17199575-17199575 on Build GRCh38
rs73876661 Chr.22: 17199416-17199416 on Build GRCh38
rs536409988 Chr.22: 17199401-17199401 on Build GRCh38
rs141781295 Chr.22: 17199445-17199445 on Build GRCh38
rs201770962 Chr.22: 17199449-17199449 on Build GRCh38
rs914094452 Chr.22: 17199484-17199484 on Build GRCh38
rs371385449 Chr.22: 17199583-17199583 on Build GRCh38
rs988265274 Chr.22: 17199474-17199474 on Build GRCh38
rs113315702 Chr.22: 17199502-17199502 on Build GRCh38

View Full Product Details