Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89191291-89191552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH15
Entrez Gene ID
1013
Gene Name
cadherin 15
Gene Aliases
CDH14, CDH3, CDHM, MCAD, MRD3
UniGene
Hs.148090
Gene Chromosome Location
Chr.16: 89171755-89195492 on Build GRCh38
Transcripts
NM_004933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247804 c.1316C>T p.P439L Chr.16: 89191413-89191413 on Build GRCh38
6315881 c.1302G>T p.Q434H Chr.16: 89191399-89191399 on Build GRCh38
5764880 c.1331T>G p.L444R Chr.16: 89191428-89191428 on Build GRCh38
106905 c.1325C>T p.P442L Chr.16: 89191422-89191422 on Build GRCh38
7076735 c.1302G>C p.Q434H Chr.16: 89191399-89191399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746239990 Chr.16: 89191349-89191349 on Build GRCh38
rs138431307 Chr.16: 89191472-89191472 on Build GRCh38
rs143389865 Chr.16: 89191372-89191372 on Build GRCh38
rs752661243 Chr.16: 89191320-89191320 on Build GRCh38
rs188795327 Chr.16: 89191333-89191333 on Build GRCh38
rs770512732 Chr.16: 89191412-89191412 on Build GRCh38
rs762540936 Chr.16: 89191370-89191370 on Build GRCh38
rs756203857 Chr.16: 89191321-89191321 on Build GRCh38
rs1030996616 Chr.16: 89191475-89191475 on Build GRCh38
rs968876589 Chr.16: 89191500-89191500 on Build GRCh38
rs944288182 Chr.16: 89191340-89191340 on Build GRCh38
rs536560983 Chr.16: 89191487-89191487 on Build GRCh38
rs760401909 Chr.16: 89191481-89191481 on Build GRCh38
rs757402303 Chr.16: 89191423-89191423 on Build GRCh38
rs921271477 Chr.16: 89191529-89191529 on Build GRCh38
rs754866953 Chr.16: 89191314-89191314 on Build GRCh38
rs771303296 Chr.16: 89191345-89191345 on Build GRCh38
rs772644150 Chr.16: 89191367-89191367 on Build GRCh38
rs759339356 Chr.16: 89191373-89191373 on Build GRCh38
rs200239402 Chr.16: 89191479-89191479 on Build GRCh38
rs140541709 Chr.16: 89191438-89191438 on Build GRCh38
rs771654189 Chr.16: 89191478-89191478 on Build GRCh38
rs755444058 Chr.16: 89191454-89191454 on Build GRCh38
rs989371827 Chr.16: 89191483-89191483 on Build GRCh38
rs767311643 Chr.16: 89191414-89191414 on Build GRCh38
rs758602653 Chr.16: 89191494-89191494 on Build GRCh38
rs151332837 Chr.16: 89191416-89191416 on Build GRCh38
rs561306828 Chr.16: 89191411-89191411 on Build GRCh38
rs771607898 Chr.16: 89191493-89191493 on Build GRCh38
rs369841519 Chr.16: 89191435-89191435 on Build GRCh38
rs757488105 Chr.16: 89191391-89191391 on Build GRCh38
rs774993875 Chr.16: 89191524-89191524 on Build GRCh38
rs373598626 Chr.16: 89191484-89191484 on Build GRCh38
rs773978015 Chr.16: 89191413-89191413 on Build GRCh38
rs373183010 Chr.16: 89191417-89191417 on Build GRCh38
rs917821092 Chr.16: 89191530-89191530 on Build GRCh38
rs879118261 Chr.16: 89191383-89191384 on Build GRCh38
rs535865456 Chr.16: 89191519-89191519 on Build GRCh38
rs753912698 Chr.16: 89191419-89191419 on Build GRCh38
rs749792180 Chr.16: 89191464-89191464 on Build GRCh38
rs771699545 Chr.16: 89191521-89191521 on Build GRCh38
rs756662636 Chr.16: 89191511-89191511 on Build GRCh38
rs372643650 Chr.16: 89191506-89191506 on Build GRCh38
rs537313663 Chr.16: 89191493-89191493 on Build GRCh38
rs569843872 Chr.16: 89191488-89191488 on Build GRCh38
rs769319278 Chr.16: 89191355-89191355 on Build GRCh38
rs80175346 Chr.16: 89191507-89191507 on Build GRCh38
rs868279691 Chr.16: 89191318-89191318 on Build GRCh38
rs777861721 Chr.16: 89191386-89191386 on Build GRCh38
rs747532135 Chr.16: 89191404-89191404 on Build GRCh38
rs201216557 Chr.16: 89191385-89191385 on Build GRCh38
rs532076741 Chr.16: 89191336-89191336 on Build GRCh38
rs951667692 Chr.16: 89191317-89191317 on Build GRCh38
rs779381603 Chr.16: 89191348-89191348 on Build GRCh38
rs1012521555 Chr.16: 89191527-89191527 on Build GRCh38
rs748710226 Chr.16: 89191462-89191462 on Build GRCh38
rs772652986 Chr.16: 89191350-89191350 on Build GRCh38
rs141704839 Chr.16: 89191422-89191422 on Build GRCh38
rs781192510 Chr.16: 89191316-89191316 on Build GRCh38
rs754003776 Chr.16: 89191390-89191390 on Build GRCh38
rs373286056 Chr.16: 89191497-89191497 on Build GRCh38
rs371395687 Chr.16: 89191351-89191351 on Build GRCh38
rs778356104 Chr.16: 89191518-89191518 on Build GRCh38
rs879757802 Chr.16: 89191482-89191482 on Build GRCh38
rs937641028 Chr.16: 89191515-89191515 on Build GRCh38
rs376123218 Chr.16: 89191453-89191453 on Build GRCh38
rs982923190 Chr.16: 89191499-89191499 on Build GRCh38
rs758601065 Chr.16: 89191436-89191436 on Build GRCh38
rs755283538 Chr.16: 89191505-89191505 on Build GRCh38
rs369285322 Chr.16: 89191346-89191346 on Build GRCh38
rs199691445 Chr.16: 89191403-89191403 on Build GRCh38
rs781671173 Chr.16: 89191456-89191456 on Build GRCh38
rs370811655 Chr.16: 89191485-89191485 on Build GRCh38
rs1016369144 Chr.16: 89191457-89191457 on Build GRCh38
rs765483021 Chr.16: 89191432-89191432 on Build GRCh38
rs770421131 Chr.16: 89191464-89191464 on Build GRCh38
rs191491461 Chr.16: 89191402-89191402 on Build GRCh38
rs866721647 Chr.16: 89191359-89191359 on Build GRCh38
rs145756506 Chr.16: 89191471-89191471 on Build GRCh38
rs772735484 Chr.16: 89191410-89191410 on Build GRCh38

View Full Product Details