Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200812271-200812518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW1
Entrez Gene ID
9689
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
BZAP45, Nbla10236
UniGene
Hs.355983
Gene Chromosome Location
Chr.2: 200811546-200823851 on Build GRCh38
Transcripts
NM_001207067.1 NM_001207069.1 NM_001321688.1 NM_001321690.1 NM_001321691.1 NM_001321693.1 NM_001321694.1 NM_014670.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6543799 c.23C>T p.P8L Chr.2: 200812430-200812430 on Build GRCh38
4154746 c.23C>G p.P8R Chr.2: 200812430-200812430 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906419502 Chr.2: 200812348-200812348 on Build GRCh38
rs780937171 Chr.2: 200812501-200812501 on Build GRCh38
rs576206260 Chr.2: 200812433-200812433 on Build GRCh38
rs562809983 Chr.2: 200812497-200812497 on Build GRCh38
rs1011995553 Chr.2: 200812298-200812298 on Build GRCh38
rs547831428 Chr.2: 200812294-200812294 on Build GRCh38
rs375146954 Chr.2: 200812422-200812422 on Build GRCh38
rs1045318889 Chr.2: 200812313-200812313 on Build GRCh38
rs896558665 Chr.2: 200812468-200812468 on Build GRCh38
rs576867353 Chr.2: 200812459-200812459 on Build GRCh38
rs906399959 Chr.2: 200812483-200812483 on Build GRCh38
rs368124923 Chr.2: 200812443-200812443 on Build GRCh38
rs1010587098 Chr.2: 200812400-200812400 on Build GRCh38
rs1044888973 Chr.2: 200812471-200812471 on Build GRCh38
rs558496103 Chr.2: 200812427-200812427 on Build GRCh38
rs953749386 Chr.2: 200812491-200812491 on Build GRCh38
rs979523941 Chr.2: 200812355-200812355 on Build GRCh38
rs937991547 Chr.2: 200812452-200812453 on Build GRCh38
rs556317029 Chr.2: 200812382-200812382 on Build GRCh38
rs560290433 Chr.2: 200812295-200812295 on Build GRCh38
rs779609519 Chr.2: 200812482-200812482 on Build GRCh38
rs1030664153 Chr.2: 200812380-200812380 on Build GRCh38
rs539587245 Chr.2: 200812447-200812447 on Build GRCh38
rs1021652630 Chr.2: 200812320-200812320 on Build GRCh38
rs986289477 Chr.2: 200812420-200812420 on Build GRCh38
rs894572151 Chr.2: 200812296-200812296 on Build GRCh38
rs372018053 Chr.2: 200812485-200812485 on Build GRCh38
rs985218118 Chr.2: 200812383-200812383 on Build GRCh38
rs767688565 Chr.2: 200812425-200812425 on Build GRCh38
rs912306342 Chr.2: 200812405-200812405 on Build GRCh38
rs953128957 Chr.2: 200812418-200812418 on Build GRCh38
rs568629348 Chr.2: 200812424-200812424 on Build GRCh38
rs1013752503 Chr.2: 200812303-200812303 on Build GRCh38
rs1031975437 Chr.2: 200812363-200812363 on Build GRCh38
rs774608455 Chr.2: 200812413-200812413 on Build GRCh38
rs544253385 Chr.2: 200812490-200812490 on Build GRCh38
rs915971512 Chr.2: 200812458-200812458 on Build GRCh38
rs919248974 Chr.2: 200812440-200812440 on Build GRCh38
rs919272925 Chr.2: 200812288-200812288 on Build GRCh38
rs966967326 Chr.2: 200812328-200812328 on Build GRCh38
rs527427974 Chr.2: 200812327-200812327 on Build GRCh38
rs1042535584 Chr.2: 200812481-200812481 on Build GRCh38
rs552290622 Chr.2: 200812373-200812373 on Build GRCh38
rs538034646 Chr.2: 200812408-200812408 on Build GRCh38

View Full Product Details