Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33581263-33581502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5800164 c.1325G>A p.R442Q Chr.21: 33581327-33581327 on Build GRCh38
5677316 c.1177A>G p.M393V Chr.21: 33581475-33581475 on Build GRCh38
1250592 c.1153C>T p.R385C Chr.21: 33581499-33581499 on Build GRCh38
1632463 c.1206G>T p.L402F Chr.21: 33581446-33581446 on Build GRCh38
1413903 c.1284G>T p.Q428H Chr.21: 33581368-33581368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937979347 Chr.21: 33581429-33581429 on Build GRCh38
rs753648048 Chr.21: 33581328-33581328 on Build GRCh38
rs765112107 Chr.21: 33581397-33581398 on Build GRCh38
rs760760475 Chr.21: 33581338-33581339 on Build GRCh38
rs766587395 Chr.21: 33581296-33581296 on Build GRCh38
rs138845591 Chr.21: 33581443-33581443 on Build GRCh38
rs993687029 Chr.21: 33581370-33581370 on Build GRCh38
rs753011971 Chr.21: 33581319-33581319 on Build GRCh38
rs138335198 Chr.21: 33581312-33581312 on Build GRCh38
rs752003766 Chr.21: 33581297-33581298 on Build GRCh38
rs747680781 Chr.21: 33581419-33581419 on Build GRCh38
rs774773262 Chr.21: 33581424-33581424 on Build GRCh38
rs771456201 Chr.21: 33581421-33581421 on Build GRCh38
rs766241167 Chr.21: 33581453-33581453 on Build GRCh38
rs547650892 Chr.21: 33581347-33581347 on Build GRCh38
rs779641743 Chr.21: 33581381-33581381 on Build GRCh38
rs764280645 Chr.21: 33581327-33581327 on Build GRCh38
rs1019172676 Chr.21: 33581350-33581350 on Build GRCh38
rs759050644 Chr.21: 33581311-33581311 on Build GRCh38
rs985261682 Chr.21: 33581404-33581404 on Build GRCh38
rs778769757 Chr.21: 33581361-33581361 on Build GRCh38
rs772599656 Chr.21: 33581446-33581446 on Build GRCh38
rs145111026 Chr.21: 33581388-33581388 on Build GRCh38
rs199603022 Chr.21: 33581385-33581385 on Build GRCh38
rs149183836 Chr.21: 33581366-33581366 on Build GRCh38
rs376502126 Chr.21: 33581295-33581295 on Build GRCh38
rs929436066 Chr.21: 33581401-33581401 on Build GRCh38
rs746684601 Chr.21: 33581387-33581387 on Build GRCh38
rs775808423 Chr.21: 33581460-33581460 on Build GRCh38
rs1009084664 Chr.21: 33581316-33581316 on Build GRCh38
rs756225695 Chr.21: 33581324-33581324 on Build GRCh38
rs758229811 Chr.21: 33581378-33581378 on Build GRCh38
rs768099763 Chr.21: 33581393-33581393 on Build GRCh38
rs776211862 Chr.21: 33581407-33581407 on Build GRCh38
rs752405565 Chr.21: 33581467-33581467 on Build GRCh38

View Full Product Details