Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49228604-49228841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX27
Entrez Gene ID
55661
Gene Name
DEAD-box helicase 27
Gene Aliases
DRS1, Drs1p, HSPC259, PP3241, RHLP, dJ686N3.1
UniGene
Hs.65234
Gene Chromosome Location
Chr.20: 49219295-49244077 on Build GRCh38
Transcripts
NM_017895.7 XM_011528888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641688 c.819C>A p.A273A Chr.20: 49228734-49228734 on Build GRCh38
6641689 c.871C>T p.R291C Chr.20: 49228786-49228786 on Build GRCh38
3547503 c.852C>T p.P284P Chr.20: 49228767-49228767 on Build GRCh38
1130613 c.855C>T p.R285R Chr.20: 49228770-49228770 on Build GRCh38
4413819 c.891C>T p.P297P Chr.20: 49228806-49228806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774124795 Chr.20: 49228669-49228669 on Build GRCh38
rs765702999 Chr.20: 49228699-49228699 on Build GRCh38
rs753592313 Chr.20: 49228686-49228686 on Build GRCh38
rs969073173 Chr.20: 49228688-49228688 on Build GRCh38
rs767426223 Chr.20: 49228675-49228675 on Build GRCh38
rs377733475 Chr.20: 49228788-49228788 on Build GRCh38
rs893740450 Chr.20: 49228661-49228661 on Build GRCh38
rs1030898166 Chr.20: 49228656-49228656 on Build GRCh38
rs144748855 Chr.20: 49228774-49228774 on Build GRCh38
rs999289620 Chr.20: 49228629-49228629 on Build GRCh38
rs759394048 Chr.20: 49228671-49228671 on Build GRCh38
rs200539083 Chr.20: 49228806-49228806 on Build GRCh38
rs747447256 Chr.20: 49228734-49228734 on Build GRCh38
rs1010664343 Chr.20: 49228673-49228673 on Build GRCh38
rs138673946 Chr.20: 49228808-49228808 on Build GRCh38
rs139155728 Chr.20: 49228708-49228708 on Build GRCh38
rs763753002 Chr.20: 49228787-49228787 on Build GRCh38
rs238165 Chr.20: 49228683-49228683 on Build GRCh38
rs761573110 Chr.20: 49228789-49228789 on Build GRCh38
rs372058226 Chr.20: 49228763-49228763 on Build GRCh38
rs775537691 Chr.20: 49228778-49228778 on Build GRCh38
rs201596939 Chr.20: 49228769-49228769 on Build GRCh38
rs756967689 Chr.20: 49228698-49228698 on Build GRCh38
rs760505992 Chr.20: 49228786-49228786 on Build GRCh38
rs238166 Chr.20: 49228725-49228725 on Build GRCh38
rs777922354 Chr.20: 49228765-49228766 on Build GRCh38
rs780574398 Chr.20: 49228726-49228726 on Build GRCh38
rs752166682 Chr.20: 49228678-49228678 on Build GRCh38
rs199775966 Chr.20: 49228700-49228700 on Build GRCh38
rs201476847 Chr.20: 49228750-49228750 on Build GRCh38
rs1018420323 Chr.20: 49228764-49228764 on Build GRCh38
rs373447678 Chr.20: 49228810-49228810 on Build GRCh38
rs750785302 Chr.20: 49228800-49228800 on Build GRCh38
rs202187886 Chr.20: 49228768-49228768 on Build GRCh38

View Full Product Details