Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100555563-100555788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
UniGene
Hs.521083
Gene Chromosome Location
Chr.7: 100539211-100568220 on Build GRCh38
Transcripts
NM_006076.4 XM_005250306.2 XM_017012112.1 XM_017012113.1 XM_017012114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3630781 c.831C>T p.L277L Chr.7: 100555689-100555689 on Build GRCh38
1699024 c.851C>T p.S284L Chr.7: 100555709-100555709 on Build GRCh38
3876331 c.752-1G>A p.? Chr.7: 100555609-100555609 on Build GRCh38
3080334 c.842G>A p.G281D Chr.7: 100555700-100555700 on Build GRCh38
6596230 c.800G>A p.S267N Chr.7: 100555658-100555658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778683068 Chr.7: 100555660-100555660 on Build GRCh38
rs375233200 Chr.7: 100555616-100555616 on Build GRCh38
rs370717204 Chr.7: 100555765-100555765 on Build GRCh38
rs773373596 Chr.7: 100555705-100555705 on Build GRCh38
rs760327205 Chr.7: 100555725-100555725 on Build GRCh38
rs201153070 Chr.7: 100555707-100555707 on Build GRCh38
rs34685790 Chr.7: 100555710-100555710 on Build GRCh38
rs753796644 Chr.7: 100555631-100555631 on Build GRCh38
rs376539393 Chr.7: 100555647-100555647 on Build GRCh38
rs1001364753 Chr.7: 100555746-100555746 on Build GRCh38
rs768979926 Chr.7: 100555676-100555676 on Build GRCh38
rs141620565 Chr.7: 100555665-100555665 on Build GRCh38
rs759818769 Chr.7: 100555714-100555714 on Build GRCh38
rs181542344 Chr.7: 100555752-100555752 on Build GRCh38
rs1041235745 Chr.7: 100555653-100555653 on Build GRCh38
rs752270655 Chr.7: 100555723-100555723 on Build GRCh38
rs767240118 Chr.7: 100555715-100555715 on Build GRCh38
rs983807528 Chr.7: 100555658-100555658 on Build GRCh38
rs773235308 Chr.7: 100555747-100555747 on Build GRCh38
rs750282208 Chr.7: 100555763-100555763 on Build GRCh38
rs757199718 Chr.7: 100555753-100555753 on Build GRCh38
rs781263256 Chr.7: 100555684-100555684 on Build GRCh38
rs748296459 Chr.7: 100555686-100555686 on Build GRCh38
rs767708937 Chr.7: 100555742-100555743 on Build GRCh38
rs867258180 Chr.7: 100555608-100555608 on Build GRCh38
rs777816840 Chr.7: 100555629-100555629 on Build GRCh38
rs769737737 Chr.7: 100555703-100555703 on Build GRCh38
rs763561479 Chr.7: 100555706-100555706 on Build GRCh38
rs778809785 Chr.7: 100555762-100555762 on Build GRCh38

View Full Product Details