Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70181027-70181252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
UniGene
Hs.720049
Gene Chromosome Location
Chr.2: 70149885-70191071 on Build GRCh38
Transcripts
NM_017880.1 XM_005264389.2 XM_005264390.2 XM_005264391.2 XM_011532937.2 XM_017004398.1 XM_017004399.1 XM_017004400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022497 c.823G>A p.G275S Chr.2: 70181163-70181163 on Build GRCh38
1669002 c.769A>G p.S257G Chr.2: 70181217-70181217 on Build GRCh38
4821673 c.804C>A p.F268L Chr.2: 70181182-70181182 on Build GRCh38
6570097 c.796T>G p.S266A Chr.2: 70181190-70181190 on Build GRCh38
4388250 c.787C>G p.Q263E Chr.2: 70181199-70181199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762199615 Chr.2: 70181143-70181143 on Build GRCh38
rs1017411996 Chr.2: 70181074-70181074 on Build GRCh38
rs763620602 Chr.2: 70181133-70181133 on Build GRCh38
rs756601319 Chr.2: 70181164-70181164 on Build GRCh38
rs977208870 Chr.2: 70181206-70181206 on Build GRCh38
rs774846597 Chr.2: 70181116-70181116 on Build GRCh38
rs992669437 Chr.2: 70181163-70181163 on Build GRCh38
rs144817179 Chr.2: 70181089-70181089 on Build GRCh38
rs564526178 Chr.2: 70181134-70181134 on Build GRCh38
rs181383847 Chr.2: 70181063-70181063 on Build GRCh38
rs753832427 Chr.2: 70181168-70181168 on Build GRCh38
rs762730554 Chr.2: 70181117-70181117 on Build GRCh38
rs1025841158 Chr.2: 70181077-70181077 on Build GRCh38
rs755274550 Chr.2: 70181196-70181196 on Build GRCh38
rs1005820327 Chr.2: 70181142-70181142 on Build GRCh38
rs148177382 Chr.2: 70181203-70181203 on Build GRCh38
rs1018264565 Chr.2: 70181145-70181145 on Build GRCh38
rs750941568 Chr.2: 70181156-70181156 on Build GRCh38
rs767807128 Chr.2: 70181152-70181152 on Build GRCh38

View Full Product Details