Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51226204-51226431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD33
Entrez Gene ID
945
Gene Name
CD33 molecule
Gene Aliases
SIGLEC-3, SIGLEC3, p67
UniGene
Hs.83731
Gene Chromosome Location
Chr.19: 51211054-51240019 on Build GRCh38
Transcripts
NM_001082618.1 NM_001177608.1 NM_001772.3 XM_011527531.2 XM_011527532.2 XM_017027508.1 XM_017027509.1 XM_017027510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3537564 c.736G>A p.D246N Chr.19: 51226347-51226347 on Build GRCh38
6298362 c.740G>T p.G247V Chr.19: 51226351-51226351 on Build GRCh38
4080579 c.733G>A p.G245R Chr.19: 51226344-51226344 on Build GRCh38
5633584 c.745+1G>A p.? Chr.19: 51226357-51226357 on Build GRCh38
999957 c.725T>C p.I242T Chr.19: 51226336-51226336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771769194 Chr.19: 51226329-51226329 on Build GRCh38
rs755698715 Chr.19: 51226402-51226403 on Build GRCh38
rs766616161 Chr.19: 51226367-51226367 on Build GRCh38
rs539344693 Chr.19: 51226395-51226395 on Build GRCh38
rs773835582 Chr.19: 51226349-51226349 on Build GRCh38
rs912847472 Chr.19: 51226331-51226331 on Build GRCh38
rs373312125 Chr.19: 51226304-51226304 on Build GRCh38
rs749130514 Chr.19: 51226394-51226394 on Build GRCh38
rs188637438 Chr.19: 51226300-51226300 on Build GRCh38
rs375460111 Chr.19: 51226279-51226279 on Build GRCh38
rs756241703 Chr.19: 51226301-51226301 on Build GRCh38
rs866924306 Chr.19: 51226354-51226354 on Build GRCh38
rs755169244 Chr.19: 51226273-51226273 on Build GRCh38
rs376681078 Chr.19: 51226305-51226305 on Build GRCh38
rs1039933048 Chr.19: 51226348-51226348 on Build GRCh38
rs774217648 Chr.19: 51226317-51226317 on Build GRCh38
rs756153648 Chr.19: 51226380-51226380 on Build GRCh38
rs971277321 Chr.19: 51226246-51226246 on Build GRCh38
rs537040950 Chr.19: 51226360-51226360 on Build GRCh38
rs781410002 Chr.19: 51226316-51226316 on Build GRCh38
rs199795598 Chr.19: 51226334-51226334 on Build GRCh38
rs755202290 Chr.19: 51226332-51226332 on Build GRCh38
rs777877198 Chr.19: 51226389-51226389 on Build GRCh38
rs1028106499 Chr.19: 51226233-51226233 on Build GRCh38
rs530607314 Chr.19: 51226254-51226254 on Build GRCh38
rs763738845 Chr.19: 51226345-51226345 on Build GRCh38
rs372484913 Chr.19: 51226263-51226263 on Build GRCh38
rs771040148 Chr.19: 51226355-51226355 on Build GRCh38
rs752980958 Chr.19: 51226379-51226379 on Build GRCh38
rs751962945 Chr.19: 51226369-51226369 on Build GRCh38
rs952089213 Chr.19: 51226276-51226276 on Build GRCh38
rs751039794 Chr.19: 51226261-51226261 on Build GRCh38
rs988337 Chr.19: 51226335-51226335 on Build GRCh38
rs781178383 Chr.19: 51226286-51226286 on Build GRCh38
rs556548617 Chr.19: 51226371-51226371 on Build GRCh38
rs372815187 Chr.19: 51226362-51226362 on Build GRCh38
rs745470324 Chr.19: 51226327-51226327 on Build GRCh38
rs751687552 Chr.19: 51226275-51226275 on Build GRCh38
rs748225001 Chr.19: 51226406-51226406 on Build GRCh38
rs373903734 Chr.19: 51226375-51226375 on Build GRCh38
rs549104922 Chr.19: 51226258-51226258 on Build GRCh38
rs368789174 Chr.19: 51226374-51226374 on Build GRCh38
rs1008804450 Chr.19: 51226393-51226393 on Build GRCh38
rs11882250 Chr.19: 51226338-51226338 on Build GRCh38

View Full Product Details