Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1656405-1656666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAMP1
Entrez Gene ID
57585
Gene Name
cramped chromatin regulator homolog 1
Gene Aliases
CRAMP1L, HN1L, TCE4
UniGene
Hs.603676
Gene Chromosome Location
Chr.16: 1614640-1677908 on Build GRCh38
Transcripts
NM_020825.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3754715 c.1816G>A p.E606K Chr.16: 1656573-1656573 on Build GRCh38
3773741 c.1668C>T p.D556D Chr.16: 1656434-1656434 on Build GRCh38
3754716 c.1807G>A p.E603K Chr.16: 1656573-1656573 on Build GRCh38
4472195 c.1757C>T p.P586L Chr.16: 1656523-1656523 on Build GRCh38
4748556 c.1889C>T p.A630V Chr.16: 1656655-1656655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61739873 Chr.16: 1656425-1656425 on Build GRCh38
rs866335522 Chr.16: 1656595-1656595 on Build GRCh38
rs80154657 Chr.16: 1656479-1656479 on Build GRCh38
rs775767622 Chr.16: 1656575-1656575 on Build GRCh38
rs1012089202 Chr.16: 1656606-1656606 on Build GRCh38
rs542745050 Chr.16: 1656496-1656496 on Build GRCh38
rs755634011 Chr.16: 1656445-1656445 on Build GRCh38
rs753456827 Chr.16: 1656607-1656607 on Build GRCh38
rs145013663 Chr.16: 1656573-1656573 on Build GRCh38
rs758649118 Chr.16: 1656430-1656430 on Build GRCh38
rs760066089 Chr.16: 1656501-1656501 on Build GRCh38
rs781721413 Chr.16: 1656618-1656618 on Build GRCh38
rs548075473 Chr.16: 1656598-1656598 on Build GRCh38
rs747772846 Chr.16: 1656557-1656557 on Build GRCh38
rs779490530 Chr.16: 1656455-1656455 on Build GRCh38
rs978745281 Chr.16: 1656623-1656623 on Build GRCh38
rs747147815 Chr.16: 1656643-1656643 on Build GRCh38
rs777091430 Chr.16: 1656564-1656564 on Build GRCh38
rs751286575 Chr.16: 1656440-1656440 on Build GRCh38
rs374460296 Chr.16: 1656626-1656626 on Build GRCh38
rs750382500 Chr.16: 1656533-1656533 on Build GRCh38
rs527950415 Chr.16: 1656591-1656591 on Build GRCh38
rs374772176 Chr.16: 1656567-1656567 on Build GRCh38
rs545390375 Chr.16: 1656544-1656544 on Build GRCh38
rs934929492 Chr.16: 1656510-1656510 on Build GRCh38
rs371731624 Chr.16: 1656594-1656594 on Build GRCh38
rs773676967 Chr.16: 1656485-1656485 on Build GRCh38
rs563031777 Chr.16: 1656515-1656515 on Build GRCh38
rs765434904 Chr.16: 1656505-1656505 on Build GRCh38
rs992663171 Chr.16: 1656624-1656624 on Build GRCh38
rs553638328 Chr.16: 1656481-1656481 on Build GRCh38
rs762046057 Chr.16: 1656589-1656589 on Build GRCh38
rs774449066 Chr.16: 1656582-1656582 on Build GRCh38
rs934564787 Chr.16: 1656499-1656499 on Build GRCh38
rs780708550 Chr.16: 1656642-1656642 on Build GRCh38
rs868767125 Chr.16: 1656457-1656457 on Build GRCh38
rs751435469 Chr.16: 1656526-1656526 on Build GRCh38
rs113314538 Chr.16: 1656524-1656524 on Build GRCh38
rs762935745 Chr.16: 1656580-1656580 on Build GRCh38
rs754452907 Chr.16: 1656608-1656608 on Build GRCh38
rs1056989413 Chr.16: 1656500-1656500 on Build GRCh38
rs573825535 Chr.16: 1656493-1656493 on Build GRCh38
rs577488354 Chr.16: 1656444-1656444 on Build GRCh38
rs746413493 Chr.16: 1656572-1656572 on Build GRCh38
rs370974722 Chr.16: 1656614-1656614 on Build GRCh38
rs758033308 Chr.16: 1656615-1656615 on Build GRCh38
rs779580535 Chr.16: 1656538-1656538 on Build GRCh38
rs767241110 Chr.16: 1656528-1656528 on Build GRCh38
rs948415333 Chr.16: 1656521-1656521 on Build GRCh38
rs757274453 Chr.16: 1656441-1656441 on Build GRCh38
rs374501457 Chr.16: 1656435-1656435 on Build GRCh38
rs915332314 Chr.16: 1656429-1656429 on Build GRCh38
rs368181087 Chr.16: 1656561-1656561 on Build GRCh38
rs766292058 Chr.16: 1656602-1656602 on Build GRCh38
rs1048724898 Chr.16: 1656542-1656542 on Build GRCh38
rs978777791 Chr.16: 1656484-1656484 on Build GRCh38
rs73499790 Chr.16: 1656539-1656539 on Build GRCh38
rs780984929 Chr.16: 1656443-1656443 on Build GRCh38
rs747324517 Chr.16: 1656490-1656490 on Build GRCh38
rs763282564 Chr.16: 1656516-1656516 on Build GRCh38
rs778428297 Chr.16: 1656553-1656553 on Build GRCh38
rs965032988 Chr.16: 1656585-1656585 on Build GRCh38
rs768712389 Chr.16: 1656581-1656581 on Build GRCh38
rs537483186 Chr.16: 1656424-1656424 on Build GRCh38
rs756006420 Chr.16: 1656534-1656534 on Build GRCh38
rs61739874 Chr.16: 1656434-1656434 on Build GRCh38
rs368776142 Chr.16: 1656461-1656461 on Build GRCh38
rs746219224 Chr.16: 1656620-1656620 on Build GRCh38

View Full Product Details