Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1243217-1243482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM132A
Entrez Gene ID
388581
Gene Name
family with sequence similarity 132 member A
Gene Aliases
C1QDC2, C1QTNF12, CTRP12
UniGene
Hs.197613
Gene Chromosome Location
Chr.1: 1242446-1247218 on Build GRCh38
Transcripts
NM_001014980.2 XM_011541435.2 XM_011541436.2 XM_011541437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354728 c.621C>G p.F207L Chr.1: 1243463-1243463 on Build GRCh38
5823948 c.635A>G p.H212R Chr.1: 1243449-1243449 on Build GRCh38
5507972 c.621C>T p.F207F Chr.1: 1243463-1243463 on Build GRCh38
6045090 c.638T>C p.V213A Chr.1: 1243446-1243446 on Build GRCh38
6259424 c.626C>A p.A209D Chr.1: 1243458-1243458 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044111245 Chr.1: 1243293-1243293 on Build GRCh38
rs987538089 Chr.1: 1243363-1243363 on Build GRCh38
rs375288834 Chr.1: 1243419-1243419 on Build GRCh38
rs10047044 Chr.1: 1243285-1243285 on Build GRCh38
rs199644687 Chr.1: 1243410-1243410 on Build GRCh38
rs899118110 Chr.1: 1243339-1243339 on Build GRCh38
rs776235459 Chr.1: 1243448-1243448 on Build GRCh38
rs113123521 Chr.1: 1243276-1243276 on Build GRCh38
rs905547264 Chr.1: 1243295-1243295 on Build GRCh38
rs377619600 Chr.1: 1243447-1243447 on Build GRCh38
rs550085611 Chr.1: 1243387-1243387 on Build GRCh38
rs986438305 Chr.1: 1243381-1243381 on Build GRCh38
rs953698911 Chr.1: 1243274-1243274 on Build GRCh38
rs924104708 Chr.1: 1243245-1243245 on Build GRCh38
rs1005163371 Chr.1: 1243318-1243318 on Build GRCh38
rs898317574 Chr.1: 1243364-1243364 on Build GRCh38
rs1028925562 Chr.1: 1243358-1243358 on Build GRCh38
rs374205352 Chr.1: 1243437-1243437 on Build GRCh38
rs1028222169 Chr.1: 1243372-1243372 on Build GRCh38
rs10047210 Chr.1: 1243286-1243286 on Build GRCh38
rs114429577 Chr.1: 1243384-1243384 on Build GRCh38
rs938417757 Chr.1: 1243300-1243300 on Build GRCh38
rs915534111 Chr.1: 1243288-1243288 on Build GRCh38
rs185313763 Chr.1: 1243256-1243256 on Build GRCh38
rs886531187 Chr.1: 1243355-1243355 on Build GRCh38
rs746299876 Chr.1: 1243409-1243409 on Build GRCh38
rs1010810038 Chr.1: 1243268-1243268 on Build GRCh38
rs563595844 Chr.1: 1243389-1243389 on Build GRCh38
rs747579965 Chr.1: 1243420-1243420 on Build GRCh38
rs1015393899 Chr.1: 1243247-1243247 on Build GRCh38
rs564107619 Chr.1: 1243370-1243370 on Build GRCh38
rs1054001028 Chr.1: 1243271-1243271 on Build GRCh38
rs540624615 Chr.1: 1243369-1243369 on Build GRCh38
rs549207399 Chr.1: 1243430-1243430 on Build GRCh38
rs996117010 Chr.1: 1243357-1243357 on Build GRCh38
rs757822660 Chr.1: 1243406-1243406 on Build GRCh38
rs561414762 Chr.1: 1243267-1243267 on Build GRCh38
rs1019202378 Chr.1: 1243399-1243399 on Build GRCh38
rs558852024 Chr.1: 1243319-1243319 on Build GRCh38
rs369025654 Chr.1: 1243436-1243436 on Build GRCh38
rs956889600 Chr.1: 1243373-1243373 on Build GRCh38
rs920961739 Chr.1: 1243269-1243269 on Build GRCh38
rs914744702 Chr.1: 1243317-1243317 on Build GRCh38
rs200025535 Chr.1: 1243445-1243445 on Build GRCh38
rs886236527 Chr.1: 1243312-1243312 on Build GRCh38
rs987977697 Chr.1: 1243242-1243242 on Build GRCh38
rs947026373 Chr.1: 1243289-1243289 on Build GRCh38
rs1044640351 Chr.1: 1243334-1243334 on Build GRCh38
rs1057294161 Chr.1: 1243310-1243310 on Build GRCh38
rs572117763 Chr.1: 1243361-1243361 on Build GRCh38
rs989679802 Chr.1: 1243382-1243382 on Build GRCh38
rs1057125497 Chr.1: 1243354-1243354 on Build GRCh38

View Full Product Details