Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 71559741-71560002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACH1
Entrez Gene ID
1602
Gene Name
dachshund family transcription factor 1
Gene Aliases
DACH
UniGene
Hs.129452
Gene Chromosome Location
Chr.13: 71437966-71868068 on Build GRCh38
Transcripts
NM_004392.6 NM_080759.5 NM_080760.5 XM_011534939.2 XM_011534940.2 XM_011534942.2 XM_017020396.1

SNP Detail

SNP ID Chromosome Location
rs199934701 Chr.13: 71559866-71559866 on Build GRCh38
rs774687803 Chr.13: 71559845-71559845 on Build GRCh38
rs1037100206 Chr.13: 71559959-71559959 on Build GRCh38
rs746257020 Chr.13: 71559811-71559811 on Build GRCh38
rs374218254 Chr.13: 71559904-71559904 on Build GRCh38
rs746180530 Chr.13: 71559911-71559911 on Build GRCh38
rs753486354 Chr.13: 71559791-71559791 on Build GRCh38
rs867568460 Chr.13: 71559946-71559946 on Build GRCh38
rs774626661 Chr.13: 71559961-71559961 on Build GRCh38
rs900891404 Chr.13: 71559947-71559947 on Build GRCh38
rs1040809407 Chr.13: 71559945-71559945 on Build GRCh38
rs767327632 Chr.13: 71559847-71559847 on Build GRCh38
rs752341288 Chr.13: 71559898-71559898 on Build GRCh38
rs749854969 Chr.13: 71559950-71559950 on Build GRCh38
rs761962203 Chr.13: 71559969-71559969 on Build GRCh38
rs534663252 Chr.13: 71559926-71559926 on Build GRCh38
rs201492130 Chr.13: 71559782-71559782 on Build GRCh38
rs764321469 Chr.13: 71559842-71559842 on Build GRCh38
rs766184140 Chr.13: 71559876-71559876 on Build GRCh38
rs939207882 Chr.13: 71559865-71559866 on Build GRCh38
rs762011848 Chr.13: 71559846-71559846 on Build GRCh38
rs568132200 Chr.13: 71559958-71559958 on Build GRCh38
rs776174060 Chr.13: 71559825-71559825 on Build GRCh38
rs369399865 Chr.13: 71559879-71559879 on Build GRCh38
rs778434084 Chr.13: 71559890-71559890 on Build GRCh38
rs377643799 Chr.13: 71559909-71559909 on Build GRCh38
rs1006642131 Chr.13: 71559923-71559923 on Build GRCh38
rs756962982 Chr.13: 71559820-71559820 on Build GRCh38
rs530403179 Chr.13: 71559805-71559805 on Build GRCh38
rs770219700 Chr.13: 71559914-71559914 on Build GRCh38
rs546719706 Chr.13: 71559928-71559928 on Build GRCh38
rs552248366 Chr.13: 71559828-71559828 on Build GRCh38
rs866262487 Chr.13: 71559968-71559968 on Build GRCh38
rs773857283 Chr.13: 71559949-71559949 on Build GRCh38
rs187346964 Chr.13: 71559785-71559785 on Build GRCh38
rs372169081 Chr.13: 71559865-71559865 on Build GRCh38
rs1053581707 Chr.13: 71559850-71559850 on Build GRCh38

View Full Product Details